Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33314790-33315063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB22
Entrez Gene ID
9278
Gene Name
zinc finger and BTB domain containing 22
Gene Aliases
BING1, ZBTB22A, ZNF297, ZNF297A, fru, fruitless
UniGene
Hs.206770
Gene Chromosome Location
Chr.6: 33314405-33317942 on Build GRCh38
Transcripts
NM_001145338.1 NM_005453.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4532257 c.1869G>A p.V623V Chr.6: 33315048-33315048 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926634840 Chr.6: 33314850-33314850 on Build GRCh38
rs748051875 Chr.6: 33314954-33314954 on Build GRCh38
rs1003885439 Chr.6: 33314867-33314867 on Build GRCh38
rs771031705 Chr.6: 33314990-33314990 on Build GRCh38
rs759499147 Chr.6: 33315003-33315003 on Build GRCh38
rs992121371 Chr.6: 33314868-33314868 on Build GRCh38
rs962636568 Chr.6: 33314832-33314832 on Build GRCh38
rs199651905 Chr.6: 33314998-33314998 on Build GRCh38
rs1043849220 Chr.6: 33314929-33314929 on Build GRCh38
rs1016793549 Chr.6: 33314824-33314824 on Build GRCh38
rs920664271 Chr.6: 33314818-33314818 on Build GRCh38
rs1061783 Chr.6: 33314851-33314851 on Build GRCh38
rs1055051634 Chr.6: 33314838-33314838 on Build GRCh38
rs899495466 Chr.6: 33314958-33314958 on Build GRCh38
rs148426865 Chr.6: 33314960-33314960 on Build GRCh38
rs771695476 Chr.6: 33314979-33314979 on Build GRCh38
rs746795718 Chr.6: 33314987-33314987 on Build GRCh38
rs532722200 Chr.6: 33314918-33314918 on Build GRCh38
rs1039728784 Chr.6: 33314948-33314948 on Build GRCh38
rs772936264 Chr.6: 33314980-33314980 on Build GRCh38
rs541696204 Chr.6: 33314941-33314941 on Build GRCh38
rs771576620 Chr.6: 33314920-33314920 on Build GRCh38
rs767769388 Chr.6: 33315029-33315029 on Build GRCh38
rs761956761 Chr.6: 33315020-33315020 on Build GRCh38
rs996986289 Chr.6: 33314846-33314846 on Build GRCh38
rs542769653 Chr.6: 33314820-33314820 on Build GRCh38
rs764112649 Chr.6: 33315006-33315006 on Build GRCh38
rs913953873 Chr.6: 33314872-33314872 on Build GRCh38
rs746477297 Chr.6: 33314826-33314826 on Build GRCh38
rs564895204 Chr.6: 33314893-33314893 on Build GRCh38
rs955022875 Chr.6: 33314855-33314855 on Build GRCh38
rs981122264 Chr.6: 33314843-33314843 on Build GRCh38
rs747749898 Chr.6: 33314916-33314916 on Build GRCh38
rs925465650 Chr.6: 33314934-33314934 on Build GRCh38
rs118050845 Chr.6: 33314819-33314819 on Build GRCh38
rs774423824 Chr.6: 33315015-33315015 on Build GRCh38
rs553607082 Chr.6: 33314971-33314971 on Build GRCh38
rs886832164 Chr.6: 33315042-33315042 on Build GRCh38

View Full Product Details