Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48029022-48029291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPZ2
Entrez Gene ID
51226
Gene Name
coatomer protein complex subunit zeta 2
Gene Aliases
zeta2-COP
UniGene
Hs.408434
Gene Chromosome Location
Chr.17: 48026167-48048091 on Build GRCh38
Transcripts
NM_016429.3 XM_011524874.2

SNP Detail

SNP ID Chromosome Location
rs571470764 Chr.17: 48029100-48029100 on Build GRCh38
rs748829543 Chr.17: 48029099-48029101 on Build GRCh38
rs745713000 Chr.17: 48029162-48029162 on Build GRCh38
rs758506977 Chr.17: 48029046-48029046 on Build GRCh38
rs897239640 Chr.17: 48029225-48029225 on Build GRCh38
rs531412051 Chr.17: 48029054-48029054 on Build GRCh38
rs9898218 Chr.17: 48029268-48029268 on Build GRCh38
rs912364263 Chr.17: 48029247-48029247 on Build GRCh38
rs373271641 Chr.17: 48029198-48029198 on Build GRCh38
rs757280373 Chr.17: 48029112-48029112 on Build GRCh38
rs769503906 Chr.17: 48029173-48029173 on Build GRCh38
rs192569571 Chr.17: 48029235-48029235 on Build GRCh38
rs944005237 Chr.17: 48029175-48029175 on Build GRCh38
rs374628919 Chr.17: 48029107-48029107 on Build GRCh38
rs185578985 Chr.17: 48029264-48029264 on Build GRCh38
rs547408770 Chr.17: 48029158-48029158 on Build GRCh38
rs918489542 Chr.17: 48029224-48029224 on Build GRCh38
rs1051662731 Chr.17: 48029152-48029152 on Build GRCh38
rs771509611 Chr.17: 48029212-48029212 on Build GRCh38
rs188355259 Chr.17: 48029176-48029176 on Build GRCh38
rs369410373 Chr.17: 48029177-48029177 on Build GRCh38
rs1049055395 Chr.17: 48029265-48029265 on Build GRCh38
rs929813969 Chr.17: 48029229-48029229 on Build GRCh38
rs986678928 Chr.17: 48029232-48029232 on Build GRCh38
rs777035951 Chr.17: 48029220-48029220 on Build GRCh38
rs750369852 Chr.17: 48029134-48029134 on Build GRCh38
rs369461034 Chr.17: 48029211-48029211 on Build GRCh38
rs768230680 Chr.17: 48029179-48029179 on Build GRCh38
rs975557823 Chr.17: 48029233-48029233 on Build GRCh38
rs146381669 Chr.17: 48029099-48029099 on Build GRCh38

View Full Product Details