Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47455826-47456097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF41
Entrez Gene ID
7592
Gene Name
zinc finger protein 41
Gene Aliases
MRX89
UniGene
Hs.496074
Gene Chromosome Location
Chr.X: 47444691-47483671 on Build GRCh38
Transcripts
NM_001324139.1 NM_001324140.1 NM_001324141.1 NM_001324142.1 NM_001324143.1 NM_001324144.1 NM_001324145.1 NM_001324147.1 NM_001324148.1 NM_001324149.1 NM_001324150.1 NM_001324151.1 NM_001324152.1 NM_001324153.1 NM_001324154.1 NM_001324155.1 NM_001324156.1 NM_001324157.1 NM_007130.3 NM_153380.3 XM_006724550.3 XM_006724555.3 XM_017029810.1 XM_017029811.1 XM_017029812.1 XM_017029813.1 XM_017029814.1 XM_017029815.1 XM_017029816.1 XM_017029817.1

SNP Detail

SNP ID Chromosome Location
rs776727494 Chr.X: 47455981-47455981 on Build GRCh38
rs770950838 Chr.X: 47455948-47455948 on Build GRCh38
rs143323743 Chr.X: 47455920-47455920 on Build GRCh38
rs951792495 Chr.X: 47455855-47455855 on Build GRCh38
rs754457805 Chr.X: 47456067-47456067 on Build GRCh38
rs772284584 Chr.X: 47455940-47455940 on Build GRCh38
rs1033440440 Chr.X: 47455990-47455990 on Build GRCh38
rs756959196 Chr.X: 47456026-47456027 on Build GRCh38
rs746383755 Chr.X: 47455936-47455936 on Build GRCh38
rs145588683 Chr.X: 47455979-47455979 on Build GRCh38
rs1045703279 Chr.X: 47456048-47456048 on Build GRCh38
rs1001571917 Chr.X: 47455986-47455986 on Build GRCh38
rs751224045 Chr.X: 47455881-47455881 on Build GRCh38
rs186650707 Chr.X: 47455906-47455906 on Build GRCh38
rs757491882 Chr.X: 47455967-47455967 on Build GRCh38
rs773026019 Chr.X: 47456026-47456026 on Build GRCh38
rs765075311 Chr.X: 47456024-47456024 on Build GRCh38
rs60428645 Chr.X: 47455984-47455984 on Build GRCh38
rs755925485 Chr.X: 47455863-47455863 on Build GRCh38
rs771308851 Chr.X: 47455929-47455929 on Build GRCh38
rs761570369 Chr.X: 47455985-47455985 on Build GRCh38
rs1010448325 Chr.X: 47456038-47456038 on Build GRCh38
rs749483903 Chr.X: 47455918-47455918 on Build GRCh38
rs754572772 Chr.X: 47455904-47455904 on Build GRCh38
rs376328166 Chr.X: 47456068-47456068 on Build GRCh38
rs779348860 Chr.X: 47455935-47455935 on Build GRCh38
rs949004207 Chr.X: 47455892-47455892 on Build GRCh38
rs762817547 Chr.X: 47456027-47456027 on Build GRCh38
rs751026477 Chr.X: 47456060-47456060 on Build GRCh38
rs765893139 Chr.X: 47456030-47456030 on Build GRCh38

View Full Product Details