Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27212135-27212408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRAID
Entrez Gene ID
51374
Gene Name
all-trans retinoic acid induced differentiation factor
Gene Aliases
APR--3, APR-3, APR3, C2orf28, HSPC013, PRO240, p18
UniGene
Hs.9527
Gene Chromosome Location
Chr.2: 27212031-27217178 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3 NM_021095.2 NR_028323.1

Gene Symbol
SLC5A6
Entrez Gene ID
8884
Gene Name
solute carrier family 5 member 6
Gene Aliases
SMVT
UniGene
Hs.435735
Gene Chromosome Location
Chr.2: 27199587-27212307 on Build GRCh38
Transcripts
NM_001170795.1 NM_016085.4 NM_080592.3 NM_021095.2 NR_028323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380133 c.194C>T p.T65M Chr.2: 27212397-27212397 on Build GRCh38
4847399 c.145G>A p.G49R Chr.2: 27212348-27212348 on Build GRCh38
7290830 c.194C>G p.T65R Chr.2: 27212397-27212397 on Build GRCh38
6608491 c.199C>A p.L67M Chr.2: 27212402-27212402 on Build GRCh38
5030722 c.193_200delACGACCCT p.T65fs*41 Chr.2: 27212396-27212403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762976723 Chr.2: 27212270-27212270 on Build GRCh38
rs369544433 Chr.2: 27212345-27212345 on Build GRCh38
rs376559066 Chr.2: 27212355-27212355 on Build GRCh38
rs780717342 Chr.2: 27212350-27212350 on Build GRCh38
rs769367297 Chr.2: 27212264-27212264 on Build GRCh38
rs868132615 Chr.2: 27212217-27212217 on Build GRCh38
rs373432775 Chr.2: 27212352-27212352 on Build GRCh38
rs767986269 Chr.2: 27212388-27212388 on Build GRCh38
rs771834206 Chr.2: 27212374-27212374 on Build GRCh38
rs11556163 Chr.2: 27212297-27212297 on Build GRCh38
rs779407253 Chr.2: 27212242-27212242 on Build GRCh38
rs753027382 Chr.2: 27212199-27212199 on Build GRCh38
rs202067119 Chr.2: 27212288-27212288 on Build GRCh38
rs200494777 Chr.2: 27212172-27212172 on Build GRCh38
rs755297948 Chr.2: 27212385-27212385 on Build GRCh38
rs751904846 Chr.2: 27212344-27212344 on Build GRCh38
rs746726954 Chr.2: 27212251-27212251 on Build GRCh38
rs1046061871 Chr.2: 27212239-27212239 on Build GRCh38
rs750988045 Chr.2: 27212301-27212301 on Build GRCh38
rs575492572 Chr.2: 27212169-27212169 on Build GRCh38
rs780860853 Chr.2: 27212255-27212255 on Build GRCh38
rs761885835 Chr.2: 27212161-27212161 on Build GRCh38
rs746919616 Chr.2: 27212253-27212254 on Build GRCh38
rs376975254 Chr.2: 27212170-27212170 on Build GRCh38
rs866493397 Chr.2: 27212244-27212244 on Build GRCh38
rs200120344 Chr.2: 27212372-27212372 on Build GRCh38
rs755750046 Chr.2: 27212354-27212354 on Build GRCh38
rs956623860 Chr.2: 27212193-27212193 on Build GRCh38
rs559815627 Chr.2: 27212331-27212331 on Build GRCh38
rs948628233 Chr.2: 27212233-27212233 on Build GRCh38
rs1007788608 Chr.2: 27212329-27212329 on Build GRCh38
rs868306306 Chr.2: 27212225-27212225 on Build GRCh38
rs1052688429 Chr.2: 27212358-27212358 on Build GRCh38
rs760793149 Chr.2: 27212205-27212205 on Build GRCh38
rs915803957 Chr.2: 27212231-27212231 on Build GRCh38
rs780271572 Chr.2: 27212247-27212247 on Build GRCh38
rs1275533 Chr.2: 27212382-27212382 on Build GRCh38
rs775983931 Chr.2: 27212155-27212155 on Build GRCh38
rs764253186 Chr.2: 27212290-27212290 on Build GRCh38
rs747680313 Chr.2: 27212260-27212260 on Build GRCh38
rs765299018 Chr.2: 27212298-27212298 on Build GRCh38
rs1019627140 Chr.2: 27212342-27212342 on Build GRCh38
rs899119256 Chr.2: 27212160-27212160 on Build GRCh38
rs995795518 Chr.2: 27212321-27212336 on Build GRCh38
rs762010882 Chr.2: 27212377-27212377 on Build GRCh38
rs757279597 Chr.2: 27212237-27212237 on Build GRCh38
rs986797180 Chr.2: 27212186-27212186 on Build GRCh38
rs912025923 Chr.2: 27212188-27212188 on Build GRCh38
rs199987825 Chr.2: 27212198-27212198 on Build GRCh38
rs908633653 Chr.2: 27212278-27212278 on Build GRCh38
rs201449113 Chr.2: 27212154-27212154 on Build GRCh38
rs774303259 Chr.2: 27212373-27212373 on Build GRCh38
rs758829508 Chr.2: 27212243-27212243 on Build GRCh38
rs867390981 Chr.2: 27212235-27212235 on Build GRCh38
rs768166971 Chr.2: 27212252-27212252 on Build GRCh38
rs1050279369 Chr.2: 27212168-27212168 on Build GRCh38
rs564260142 Chr.2: 27212275-27212275 on Build GRCh38
rs762557057 Chr.2: 27212295-27212295 on Build GRCh38
rs765352518 Chr.2: 27212378-27212378 on Build GRCh38
rs1027955698 Chr.2: 27212316-27212323 on Build GRCh38
rs768222803 Chr.2: 27212286-27212287 on Build GRCh38
rs773247722 Chr.2: 27212381-27212381 on Build GRCh38
rs1030750923 Chr.2: 27212184-27212184 on Build GRCh38
rs907008953 Chr.2: 27212246-27212246 on Build GRCh38
rs201210592 Chr.2: 27212318-27212318 on Build GRCh38
rs775098924 Chr.2: 27212268-27212268 on Build GRCh38
rs944561840 Chr.2: 27212346-27212346 on Build GRCh38
rs919368775 Chr.2: 27212209-27212209 on Build GRCh38
rs752042458 Chr.2: 27212384-27212384 on Build GRCh38
rs753882664 Chr.2: 27212232-27212232 on Build GRCh38
rs969637389 Chr.2: 27212218-27212218 on Build GRCh38
rs1010606517 Chr.2: 27212197-27212197 on Build GRCh38
rs769613846 Chr.2: 27212227-27212227 on Build GRCh38
rs776906751 Chr.2: 27212375-27212375 on Build GRCh38
rs776777155 Chr.2: 27212292-27212292 on Build GRCh38
rs201195587 Chr.2: 27212228-27212228 on Build GRCh38
rs369868002 Chr.2: 27212324-27212324 on Build GRCh38
rs749286000 Chr.2: 27212360-27212360 on Build GRCh38

View Full Product Details