Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56351279-56351536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASDH
Entrez Gene ID
132949
Gene Name
aminoadipate-semialdehyde dehydrogenase
Gene Aliases
ACSF4, LYS2, NRPS1098, NRPS998
UniGene
Hs.104347
Gene Chromosome Location
Chr.4: 56337687-56387677 on Build GRCh38
Transcripts
NM_001286668.1 NM_001286669.1 NM_001286670.1 NM_001286671.1 NM_001286672.1 NM_001323890.1 NM_001323892.1 NM_001323893.1 NM_001323899.1 NM_181806.3 XM_017007740.1 XM_017007741.1 XM_017007742.1 XM_017007743.1 XM_017007744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826017 c.1619A>G p.Y540C Chr.4: 56351415-56351415 on Build GRCh38
6100786 c.1577-1G>A p.? Chr.4: 56351458-56351458 on Build GRCh38
3302031 c.1675T>A p.L559I Chr.4: 56351359-56351359 on Build GRCh38
3302032 c.1621A>G p.I541V Chr.4: 56351413-56351413 on Build GRCh38
3826018 c.1619A>G p.Y540C Chr.4: 56351415-56351415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762038937 Chr.4: 56351426-56351426 on Build GRCh38
rs759579318 Chr.4: 56351377-56351377 on Build GRCh38
rs751706495 Chr.4: 56351398-56351398 on Build GRCh38
rs772807710 Chr.4: 56351312-56351312 on Build GRCh38
rs764207178 Chr.4: 56351392-56351392 on Build GRCh38
rs766675210 Chr.4: 56351472-56351472 on Build GRCh38
rs755479500 Chr.4: 56351502-56351502 on Build GRCh38
rs754300647 Chr.4: 56351474-56351474 on Build GRCh38
rs1041200757 Chr.4: 56351306-56351306 on Build GRCh38
rs776664368 Chr.4: 56351364-56351364 on Build GRCh38
rs750648442 Chr.4: 56351462-56351462 on Build GRCh38
rs11727715 Chr.4: 56351437-56351437 on Build GRCh38
rs770682839 Chr.4: 56351332-56351332 on Build GRCh38
rs1032694635 Chr.4: 56351413-56351413 on Build GRCh38
rs944930089 Chr.4: 56351359-56351359 on Build GRCh38
rs901330981 Chr.4: 56351351-56351351 on Build GRCh38
rs900254397 Chr.4: 56351376-56351376 on Build GRCh38
rs747906678 Chr.4: 56351373-56351374 on Build GRCh38
rs561817414 Chr.4: 56351367-56351367 on Build GRCh38
rs572364329 Chr.4: 56351467-56351467 on Build GRCh38
rs879167447 Chr.4: 56351403-56351403 on Build GRCh38
rs776896059 Chr.4: 56351310-56351312 on Build GRCh38
rs760488890 Chr.4: 56351326-56351326 on Build GRCh38

View Full Product Details