Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 33406831-33407074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT10
Entrez Gene ID
341359
Gene Name
synaptotagmin 10
Gene Aliases
-
UniGene
Hs.118703
Gene Chromosome Location
Chr.12: 33375413-33439819 on Build GRCh38
Transcripts
NM_198992.3 XM_011520644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2206184 c.1013T>C p.L338S Chr.12: 33406853-33406853 on Build GRCh38
6072435 c.908C>A p.P303H Chr.12: 33406958-33406958 on Build GRCh38
6553130 c.866G>A p.R289K Chr.12: 33407000-33407000 on Build GRCh38
6531473 c.896C>G p.T299S Chr.12: 33406970-33406970 on Build GRCh38
4752434 c.960T>G p.D320E Chr.12: 33406906-33406906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758611055 Chr.12: 33406900-33406900 on Build GRCh38
rs761826830 Chr.12: 33406883-33406883 on Build GRCh38
rs751127584 Chr.12: 33406961-33406961 on Build GRCh38
rs754823879 Chr.12: 33406916-33406916 on Build GRCh38
rs752724042 Chr.12: 33406931-33406931 on Build GRCh38
rs767787877 Chr.12: 33406887-33406887 on Build GRCh38
rs759382079 Chr.12: 33406944-33406944 on Build GRCh38
rs1042570996 Chr.12: 33406996-33406996 on Build GRCh38
rs762776606 Chr.12: 33407044-33407044 on Build GRCh38
rs767336073 Chr.12: 33407008-33407008 on Build GRCh38
rs745458244 Chr.12: 33406984-33406984 on Build GRCh38
rs765148812 Chr.12: 33407035-33407035 on Build GRCh38
rs756761234 Chr.12: 33406963-33406963 on Build GRCh38
rs780248794 Chr.12: 33407005-33407005 on Build GRCh38
rs541132224 Chr.12: 33407010-33407010 on Build GRCh38
rs752525618 Chr.12: 33407039-33407039 on Build GRCh38
rs770240151 Chr.12: 33407004-33407004 on Build GRCh38
rs780107776 Chr.12: 33406960-33406960 on Build GRCh38
rs146031713 Chr.12: 33407009-33407009 on Build GRCh38
rs912954634 Chr.12: 33406934-33406934 on Build GRCh38
rs534242250 Chr.12: 33406932-33406932 on Build GRCh38
rs764919133 Chr.12: 33406924-33406924 on Build GRCh38
rs769167593 Chr.12: 33407007-33407007 on Build GRCh38
rs373995269 Chr.12: 33406865-33406865 on Build GRCh38
rs760778883 Chr.12: 33407029-33407029 on Build GRCh38
rs773533809 Chr.12: 33407012-33407012 on Build GRCh38
rs763243494 Chr.12: 33407002-33407002 on Build GRCh38
rs747135559 Chr.12: 33406997-33406997 on Build GRCh38
rs866101269 Chr.12: 33406959-33406959 on Build GRCh38
rs144188198 Chr.12: 33406979-33406979 on Build GRCh38
rs141819559 Chr.12: 33406912-33406912 on Build GRCh38
rs777482060 Chr.12: 33406953-33406953 on Build GRCh38

View Full Product Details