Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1294393-1294634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
UniGene
Hs.535257
Gene Chromosome Location
Chr.1: 1292384-1307925 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5509648 c.1835C>T p.A612V Chr.1: 1294481-1294481 on Build GRCh38
1181546 c.2048C>T p.A683V Chr.1: 1294493-1294493 on Build GRCh38
1181547 c.1823C>T p.A608V Chr.1: 1294493-1294493 on Build GRCh38
5509646 c.1872G>A p.A624A Chr.1: 1294444-1294444 on Build GRCh38
4476133 c.1820C>T p.P607L Chr.1: 1294496-1294496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374826827 Chr.1: 1294584-1294584 on Build GRCh38
rs760466206 Chr.1: 1294528-1294528 on Build GRCh38
rs759645640 Chr.1: 1294494-1294494 on Build GRCh38
rs775697872 Chr.1: 1294506-1294506 on Build GRCh38
rs778368288 Chr.1: 1294473-1294473 on Build GRCh38
rs867511621 Chr.1: 1294535-1294535 on Build GRCh38
rs776724403 Chr.1: 1294414-1294414 on Build GRCh38
rs111875048 Chr.1: 1294490-1294490 on Build GRCh38
rs750226138 Chr.1: 1294461-1294461 on Build GRCh38
rs747334355 Chr.1: 1294486-1294486 on Build GRCh38
rs866469268 Chr.1: 1294575-1294575 on Build GRCh38
rs754605511 Chr.1: 1294540-1294540 on Build GRCh38
rs867514004 Chr.1: 1294464-1294464 on Build GRCh38
rs747708021 Chr.1: 1294479-1294479 on Build GRCh38
rs759603957 Chr.1: 1294434-1294434 on Build GRCh38
rs752863196 Chr.1: 1294445-1294445 on Build GRCh38
rs907571116 Chr.1: 1294576-1294576 on Build GRCh38
rs768874810 Chr.1: 1294607-1294607 on Build GRCh38
rs1053375951 Chr.1: 1294558-1294558 on Build GRCh38
rs574689077 Chr.1: 1294573-1294573 on Build GRCh38
rs867918836 Chr.1: 1294612-1294612 on Build GRCh38
rs868832637 Chr.1: 1294596-1294596 on Build GRCh38
rs752200309 Chr.1: 1294553-1294553 on Build GRCh38
rs921358604 Chr.1: 1294491-1294491 on Build GRCh38
rs367654678 Chr.1: 1294615-1294615 on Build GRCh38
rs766122762 Chr.1: 1294537-1294537 on Build GRCh38
rs988021610 Chr.1: 1294475-1294486 on Build GRCh38
rs771214380 Chr.1: 1294487-1294487 on Build GRCh38
rs763066539 Chr.1: 1294587-1294587 on Build GRCh38
rs866644774 Chr.1: 1294579-1294579 on Build GRCh38
rs763189337 Chr.1: 1294512-1294512 on Build GRCh38
rs778285678 Chr.1: 1294559-1294559 on Build GRCh38
rs865803831 Chr.1: 1294602-1294602 on Build GRCh38
rs748911098 Chr.1: 1294471-1294471 on Build GRCh38
rs770134377 Chr.1: 1294582-1294582 on Build GRCh38
rs776871542 Chr.1: 1294493-1294493 on Build GRCh38
rs773775845 Chr.1: 1294483-1294483 on Build GRCh38
rs771507543 Chr.1: 1294571-1294571 on Build GRCh38
rs773157387 Chr.1: 1294610-1294610 on Build GRCh38
rs778597599 Chr.1: 1294548-1294548 on Build GRCh38
rs750337212 Chr.1: 1294519-1294519 on Build GRCh38
rs540738244 Chr.1: 1294498-1294498 on Build GRCh38
rs202162654 Chr.1: 1294480-1294480 on Build GRCh38
rs932805838 Chr.1: 1294497-1294497 on Build GRCh38
rs989999025 Chr.1: 1294481-1294481 on Build GRCh38
rs866709468 Chr.1: 1294590-1294590 on Build GRCh38
rs753712050 Chr.1: 1294538-1294538 on Build GRCh38
rs757910950 Chr.1: 1294554-1294554 on Build GRCh38
rs760502036 Chr.1: 1294611-1294611 on Build GRCh38
rs200117221 Chr.1: 1294465-1294465 on Build GRCh38
rs200710534 Chr.1: 1294566-1294566 on Build GRCh38
rs371863796 Chr.1: 1294581-1294581 on Build GRCh38
rs368485636 Chr.1: 1294446-1294446 on Build GRCh38
rs767619464 Chr.1: 1294453-1294453 on Build GRCh38
rs767493080 Chr.1: 1294517-1294517 on Build GRCh38
rs558704852 Chr.1: 1294444-1294444 on Build GRCh38

View Full Product Details