Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61132306-61132569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS37C
Entrez Gene ID
55048
Gene Name
VPS37C, ESCRT-I subunit
Gene Aliases
-
UniGene
Hs.523715
Gene Chromosome Location
Chr.11: 61130256-61161444 on Build GRCh38
Transcripts
NM_017966.4 XM_005274077.3 XM_017017955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188257 c.460C>T p.Q154* Chr.11: 61132428-61132428 on Build GRCh38
6537379 c.463G>C p.E155Q Chr.11: 61132425-61132425 on Build GRCh38
1188258 c.448G>A p.V150M Chr.11: 61132440-61132440 on Build GRCh38
6552173 c.423G>A p.M141I Chr.11: 61132465-61132465 on Build GRCh38
6552174 c.399G>A p.L133L Chr.11: 61132489-61132489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989935799 Chr.11: 61132446-61132446 on Build GRCh38
rs746287465 Chr.11: 61132368-61132368 on Build GRCh38
rs375223987 Chr.11: 61132367-61132367 on Build GRCh38
rs369483972 Chr.11: 61132371-61132371 on Build GRCh38
rs774129727 Chr.11: 61132417-61132417 on Build GRCh38
rs760267596 Chr.11: 61132513-61132513 on Build GRCh38
rs199606009 Chr.11: 61132355-61132355 on Build GRCh38
rs369758363 Chr.11: 61132496-61132496 on Build GRCh38
rs72914704 Chr.11: 61132546-61132546 on Build GRCh38
rs147453885 Chr.11: 61132442-61132442 on Build GRCh38
rs752255481 Chr.11: 61132438-61132438 on Build GRCh38
rs561266642 Chr.11: 61132461-61132461 on Build GRCh38
rs560333251 Chr.11: 61132360-61132360 on Build GRCh38
rs934501557 Chr.11: 61132474-61132474 on Build GRCh38
rs755306619 Chr.11: 61132449-61132449 on Build GRCh38
rs767195781 Chr.11: 61132495-61132495 on Build GRCh38
rs200081898 Chr.11: 61132455-61132455 on Build GRCh38
rs558890134 Chr.11: 61132387-61132387 on Build GRCh38
rs766094819 Chr.11: 61132346-61132346 on Build GRCh38
rs755593200 Chr.11: 61132440-61132440 on Build GRCh38
rs769900068 Chr.11: 61132397-61132397 on Build GRCh38
rs373025668 Chr.11: 61132384-61132384 on Build GRCh38
rs201478286 Chr.11: 61132443-61132443 on Build GRCh38
rs748222725 Chr.11: 61132389-61132389 on Build GRCh38
rs774073165 Chr.11: 61132486-61132486 on Build GRCh38
rs2232142 Chr.11: 61132343-61132343 on Build GRCh38
rs199507258 Chr.11: 61132350-61132350 on Build GRCh38
rs761843001 Chr.11: 61132334-61132334 on Build GRCh38
rs768929755 Chr.11: 61132386-61132386 on Build GRCh38
rs149358909 Chr.11: 61132441-61132441 on Build GRCh38
rs759251755 Chr.11: 61132425-61132425 on Build GRCh38
rs781540368 Chr.11: 61132451-61132451 on Build GRCh38
rs763439902 Chr.11: 61132522-61132522 on Build GRCh38
rs955654233 Chr.11: 61132377-61132377 on Build GRCh38
rs766110441 Chr.11: 61132408-61132408 on Build GRCh38
rs940024093 Chr.11: 61132512-61132512 on Build GRCh38
rs759302652 Chr.11: 61132489-61132489 on Build GRCh38
rs867212254 Chr.11: 61132543-61132543 on Build GRCh38
rs772991189 Chr.11: 61132398-61132398 on Build GRCh38
rs753326154 Chr.11: 61132521-61132521 on Build GRCh38
rs751202506 Chr.11: 61132349-61132349 on Build GRCh38
rs377533753 Chr.11: 61132452-61132452 on Build GRCh38
rs140024820 Chr.11: 61132457-61132457 on Build GRCh38
rs556654929 Chr.11: 61132344-61132344 on Build GRCh38
rs770898116 Chr.11: 61132467-61132467 on Build GRCh38
rs763521473 Chr.11: 61132519-61132519 on Build GRCh38
rs762906858 Chr.11: 61132406-61132406 on Build GRCh38
rs777019135 Chr.11: 61132521-61132521 on Build GRCh38
rs767145002 Chr.11: 61132426-61132426 on Build GRCh38
rs117231827 Chr.11: 61132460-61132460 on Build GRCh38
rs574643931 Chr.11: 61132472-61132472 on Build GRCh38
rs369574233 Chr.11: 61132342-61132342 on Build GRCh38
rs767020079 Chr.11: 61132354-61132354 on Build GRCh38
rs765259626 Chr.11: 61132339-61132339 on Build GRCh38
rs757821185 Chr.11: 61132356-61132356 on Build GRCh38

View Full Product Details