Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36039506-36039771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101927572
Entrez Gene ID
101927572
Gene Name
uncharacterized LOC101927572
Gene Aliases
-
UniGene
Hs.676116
Gene Chromosome Location
Chr.19: 36014499-36045997 on Build GRCh38
Transcripts
NM_001290056.1 NM_152658.2 XM_005258623.2 XM_011526604.2

Gene Symbol
THAP8
Entrez Gene ID
199745
Gene Name
THAP domain containing 8
Gene Aliases
-
UniGene
Hs.350209
Gene Chromosome Location
Chr.19: 36034984-36054785 on Build GRCh38
Transcripts
NM_001290056.1 NM_152658.2 XM_005258623.2 XM_011526604.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3716499 c.318G>A p.P106P Chr.19: 36039677-36039677 on Build GRCh38
5445693 c.335A>G p.K112R Chr.19: 36039660-36039660 on Build GRCh38
995354 c.462G>A p.A154A Chr.19: 36039533-36039533 on Build GRCh38
7265980 c.476G>A p.R159Q Chr.19: 36039519-36039519 on Build GRCh38
6289959 c.403C>T p.P135S Chr.19: 36039592-36039592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546292888 Chr.19: 36039711-36039711 on Build GRCh38
rs745636819 Chr.19: 36039605-36039605 on Build GRCh38
rs34250145 Chr.19: 36039526-36039526 on Build GRCh38
rs188124732 Chr.19: 36039545-36039545 on Build GRCh38
rs997419789 Chr.19: 36039632-36039632 on Build GRCh38
rs993120241 Chr.19: 36039706-36039706 on Build GRCh38
rs903652768 Chr.19: 36039594-36039594 on Build GRCh38
rs762840041 Chr.19: 36039654-36039654 on Build GRCh38
rs368095406 Chr.19: 36039674-36039674 on Build GRCh38
rs753131009 Chr.19: 36039702-36039702 on Build GRCh38
rs770987762 Chr.19: 36039585-36039585 on Build GRCh38
rs776977832 Chr.19: 36039602-36039602 on Build GRCh38
rs917581713 Chr.19: 36039721-36039721 on Build GRCh38
rs374386221 Chr.19: 36039750-36039750 on Build GRCh38
rs746140437 Chr.19: 36039751-36039751 on Build GRCh38
rs1051278506 Chr.19: 36039680-36039680 on Build GRCh38
rs762261158 Chr.19: 36039624-36039624 on Build GRCh38
rs765676732 Chr.19: 36039692-36039692 on Build GRCh38
rs564576286 Chr.19: 36039712-36039712 on Build GRCh38
rs75451634 Chr.19: 36039528-36039528 on Build GRCh38
rs941787388 Chr.19: 36039658-36039658 on Build GRCh38
rs999296119 Chr.19: 36039536-36039536 on Build GRCh38
rs371566939 Chr.19: 36039578-36039578 on Build GRCh38
rs772137578 Chr.19: 36039552-36039552 on Build GRCh38
rs945033442 Chr.19: 36039740-36039740 on Build GRCh38
rs367994008 Chr.19: 36039677-36039677 on Build GRCh38
rs772421009 Chr.19: 36039686-36039687 on Build GRCh38
rs772249872 Chr.19: 36039572-36039572 on Build GRCh38
rs3810450 Chr.19: 36039660-36039660 on Build GRCh38
rs757415599 Chr.19: 36039739-36039739 on Build GRCh38
rs202180130 Chr.19: 36039681-36039681 on Build GRCh38
rs777948106 Chr.19: 36039576-36039576 on Build GRCh38
rs1030273427 Chr.19: 36039638-36039638 on Build GRCh38
rs374686785 Chr.19: 36039612-36039612 on Build GRCh38
rs755441909 Chr.19: 36039535-36039535 on Build GRCh38
rs928987399 Chr.19: 36039610-36039610 on Build GRCh38
rs980689276 Chr.19: 36039678-36039678 on Build GRCh38
rs375640594 Chr.19: 36039675-36039675 on Build GRCh38
rs957715994 Chr.19: 36039547-36039547 on Build GRCh38
rs775257799 Chr.19: 36039641-36039641 on Build GRCh38
rs915766666 Chr.19: 36039577-36039577 on Build GRCh38
rs945105105 Chr.19: 36039584-36039584 on Build GRCh38
rs192898190 Chr.19: 36039730-36039730 on Build GRCh38
rs927554662 Chr.19: 36039544-36039544 on Build GRCh38
rs113554294 Chr.19: 36039534-36039534 on Build GRCh38
rs201068048 Chr.19: 36039703-36039703 on Build GRCh38
rs989954154 Chr.19: 36039561-36039561 on Build GRCh38
rs774964986 Chr.19: 36039543-36039543 on Build GRCh38
rs773713000 Chr.19: 36039728-36039728 on Build GRCh38

View Full Product Details