Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37996280-37996553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTI1
Entrez Gene ID
9675
Gene Name
TELO2 interacting protein 1
Gene Aliases
KIAA0406, smg-10
UniGene
Hs.655481
Gene Chromosome Location
Chr.20: 37983007-38033468 on Build GRCh38
Transcripts
NM_001303457.1 NM_014657.2 XM_011529114.1 XM_017028148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3546083 c.3043C>T p.L1015F Chr.20: 37996418-37996418 on Build GRCh38
321172 c.3044T>A p.L1015H Chr.20: 37996417-37996417 on Build GRCh38
6332859 c.3081C>A p.A1027A Chr.20: 37996380-37996380 on Build GRCh38
2760827 c.3029C>T p.A1010V Chr.20: 37996432-37996432 on Build GRCh38
6421458 c.3023C>T p.A1008V Chr.20: 37996438-37996438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370819283 Chr.20: 37996325-37996325 on Build GRCh38
rs747245849 Chr.20: 37996503-37996503 on Build GRCh38
rs776830355 Chr.20: 37996512-37996512 on Build GRCh38
rs368305458 Chr.20: 37996342-37996342 on Build GRCh38
rs777673967 Chr.20: 37996343-37996343 on Build GRCh38
rs769011951 Chr.20: 37996382-37996382 on Build GRCh38
rs772864335 Chr.20: 37996479-37996479 on Build GRCh38
rs147441837 Chr.20: 37996326-37996326 on Build GRCh38
rs572537626 Chr.20: 37996379-37996379 on Build GRCh38
rs1053119026 Chr.20: 37996355-37996355 on Build GRCh38
rs766890800 Chr.20: 37996408-37996408 on Build GRCh38
rs778954246 Chr.20: 37996484-37996484 on Build GRCh38
rs200671578 Chr.20: 37996374-37996374 on Build GRCh38
rs140168073 Chr.20: 37996401-37996401 on Build GRCh38
rs755764274 Chr.20: 37996419-37996419 on Build GRCh38
rs954387122 Chr.20: 37996511-37996511 on Build GRCh38
rs749246925 Chr.20: 37996344-37996344 on Build GRCh38
rs758534448 Chr.20: 37996500-37996500 on Build GRCh38
rs768923986 Chr.20: 37996507-37996507 on Build GRCh38
rs34900517 Chr.20: 37996378-37996378 on Build GRCh38
rs780039217 Chr.20: 37996502-37996502 on Build GRCh38
rs752431839 Chr.20: 37996415-37996415 on Build GRCh38
rs775708714 Chr.20: 37996371-37996371 on Build GRCh38
rs2273349 Chr.20: 37996354-37996354 on Build GRCh38
rs41296227 Chr.20: 37996527-37996527 on Build GRCh38
rs772019969 Chr.20: 37996370-37996370 on Build GRCh38
rs763998717 Chr.20: 37996429-37996429 on Build GRCh38
rs767241856 Chr.20: 37996336-37996336 on Build GRCh38
rs535684714 Chr.20: 37996443-37996443 on Build GRCh38
rs765840379 Chr.20: 37996400-37996400 on Build GRCh38
rs756968196 Chr.20: 37996483-37996483 on Build GRCh38
rs983231078 Chr.20: 37996516-37996516 on Build GRCh38
rs762070986 Chr.20: 37996399-37996399 on Build GRCh38
rs752376612 Chr.20: 37996338-37996338 on Build GRCh38
rs745996457 Chr.20: 37996368-37996368 on Build GRCh38
rs753748379 Chr.20: 37996463-37996463 on Build GRCh38
rs776922927 Chr.20: 37996385-37996385 on Build GRCh38

View Full Product Details