Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56516807-56517080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT7
Entrez Gene ID
140687
Gene Name
glucosaminyl (N-acetyl) transferase family member 7
Gene Aliases
C20orf105, dJ1153D9.2, gcnt
UniGene
Hs.655916
Gene Chromosome Location
Chr.20: 56491492-56525925 on Build GRCh38
Transcripts
NM_080615.1 NM_001283035.1 NM_001283036.1 NM_001283037.1 NM_016407.4 XM_017027872.1

Gene Symbol
RTFDC1
Entrez Gene ID
51507
Gene Name
replication termination factor 2 domain containing 1
Gene Aliases
C20orf43, CDAO5, HSPC164, SHUJUN-3
UniGene
Hs.517134
Gene Chromosome Location
Chr.20: 56468585-56518886 on Build GRCh38
Transcripts
NM_080615.1 NM_001283035.1 NM_001283036.1 NM_001283037.1 NM_016407.4 XM_017027872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5730591 c.709T>A p.S237T Chr.20: 56516962-56516962 on Build GRCh38
7153585 c.632C>A p.P211Q Chr.20: 56516975-56516975 on Build GRCh38
3707859 c.728T>G p.V243G Chr.20: 56516981-56516981 on Build GRCh38
7153584 c.722C>A p.P241Q Chr.20: 56516975-56516975 on Build GRCh38
3707858 c.638T>G p.V213G Chr.20: 56516981-56516981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028179411 Chr.20: 56517008-56517008 on Build GRCh38
rs776125741 Chr.20: 56516938-56516938 on Build GRCh38
rs749171282 Chr.20: 56516899-56516899 on Build GRCh38
rs535688127 Chr.20: 56516880-56516880 on Build GRCh38
rs762300777 Chr.20: 56517049-56517049 on Build GRCh38
rs763348585 Chr.20: 56516987-56516987 on Build GRCh38
rs775067666 Chr.20: 56516922-56516922 on Build GRCh38
rs554735673 Chr.20: 56516855-56516855 on Build GRCh38
rs900807932 Chr.20: 56516950-56516950 on Build GRCh38
rs534734550 Chr.20: 56516843-56516846 on Build GRCh38
rs368029194 Chr.20: 56516985-56516985 on Build GRCh38
rs1056002658 Chr.20: 56517042-56517042 on Build GRCh38
rs765757381 Chr.20: 56516984-56516984 on Build GRCh38
rs777631410 Chr.20: 56516987-56516987 on Build GRCh38
rs906987202 Chr.20: 56516861-56516862 on Build GRCh38
rs998972227 Chr.20: 56516883-56516883 on Build GRCh38
rs200847767 Chr.20: 56516907-56516907 on Build GRCh38
rs919572808 Chr.20: 56516851-56516851 on Build GRCh38
rs757962749 Chr.20: 56516921-56516922 on Build GRCh38
rs775769652 Chr.20: 56516921-56516921 on Build GRCh38
rs368600068 Chr.20: 56517052-56517052 on Build GRCh38
rs764736564 Chr.20: 56517027-56517027 on Build GRCh38
rs767997373 Chr.20: 56516934-56516934 on Build GRCh38
rs534046428 Chr.20: 56516919-56516919 on Build GRCh38
rs779032120 Chr.20: 56517041-56517041 on Build GRCh38
rs766620502 Chr.20: 56516988-56516988 on Build GRCh38
rs762270957 Chr.20: 56516974-56516974 on Build GRCh38
rs565500124 Chr.20: 56516916-56516916 on Build GRCh38
rs374689336 Chr.20: 56516923-56516923 on Build GRCh38
rs184683537 Chr.20: 56516853-56516853 on Build GRCh38
rs368137777 Chr.20: 56516881-56516881 on Build GRCh38
rs757235236 Chr.20: 56517034-56517034 on Build GRCh38
rs376559631 Chr.20: 56517032-56517032 on Build GRCh38
rs751924997 Chr.20: 56516892-56516892 on Build GRCh38
rs576859477 Chr.20: 56517028-56517028 on Build GRCh38
rs774236085 Chr.20: 56516914-56516914 on Build GRCh38
rs750145465 Chr.20: 56517019-56517020 on Build GRCh38
rs952578113 Chr.20: 56517024-56517024 on Build GRCh38
rs879246159 Chr.20: 56517012-56517012 on Build GRCh38
rs938447324 Chr.20: 56517015-56517015 on Build GRCh38
rs752298654 Chr.20: 56516920-56516923 on Build GRCh38
rs200348126 Chr.20: 56517007-56517007 on Build GRCh38
rs894669762 Chr.20: 56516835-56516835 on Build GRCh38

View Full Product Details