Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37537451-37537704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF793
Entrez Gene ID
390927
Gene Name
zinc finger protein 793
Gene Aliases
-
UniGene
Hs.568010
Gene Chromosome Location
Chr.19: 37506939-37543337 on Build GRCh38
Transcripts
NM_001013659.2 XM_005258927.3 XM_006723213.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077586 c.873C>T p.A291A Chr.19: 37537531-37537531 on Build GRCh38
4077587 c.942T>G p.S314R Chr.19: 37537600-37537600 on Build GRCh38
7270599 c.1019G>A p.R340H Chr.19: 37537677-37537677 on Build GRCh38
1271798 c.854_855insT p.C288fs*28 Chr.19: 37537512-37537513 on Build GRCh38
5758653 c.906G>T p.Q302H Chr.19: 37537564-37537564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878910993 Chr.19: 37537503-37537503 on Build GRCh38
rs765809832 Chr.19: 37537530-37537530 on Build GRCh38
rs139014237 Chr.19: 37537606-37537606 on Build GRCh38
rs923285660 Chr.19: 37537566-37537566 on Build GRCh38
rs757499893 Chr.19: 37537635-37537635 on Build GRCh38
rs368936802 Chr.19: 37537536-37537536 on Build GRCh38
rs373734998 Chr.19: 37537638-37537638 on Build GRCh38
rs201336836 Chr.19: 37537550-37537550 on Build GRCh38
rs377400755 Chr.19: 37537647-37537647 on Build GRCh38
rs866571724 Chr.19: 37537503-37537504 on Build GRCh38
rs766943226 Chr.19: 37537512-37537513 on Build GRCh38
rs751784572 Chr.19: 37537551-37537551 on Build GRCh38
rs774206700 Chr.19: 37537491-37537491 on Build GRCh38
rs914096642 Chr.19: 37537483-37537483 on Build GRCh38
rs188659958 Chr.19: 37537582-37537582 on Build GRCh38
rs192785678 Chr.19: 37537614-37537614 on Build GRCh38
rs1012862478 Chr.19: 37537511-37537511 on Build GRCh38
rs773935686 Chr.19: 37537601-37537601 on Build GRCh38
rs375572663 Chr.19: 37537671-37537671 on Build GRCh38
rs776998243 Chr.19: 37537528-37537528 on Build GRCh38
rs761556659 Chr.19: 37537521-37537521 on Build GRCh38
rs374828147 Chr.19: 37537516-37537516 on Build GRCh38
rs751883374 Chr.19: 37537676-37537676 on Build GRCh38
rs768259830 Chr.19: 37537490-37537490 on Build GRCh38
rs780870370 Chr.19: 37537570-37537570 on Build GRCh38
rs748871202 Chr.19: 37537484-37537484 on Build GRCh38
rs772705783 Chr.19: 37537607-37537607 on Build GRCh38
rs751588336 Chr.19: 37537633-37537633 on Build GRCh38
rs375746014 Chr.19: 37537572-37537572 on Build GRCh38
rs45505797 Chr.19: 37537677-37537677 on Build GRCh38
rs759991989 Chr.19: 37537612-37537612 on Build GRCh38
rs373213372 Chr.19: 37537592-37537592 on Build GRCh38
rs779709556 Chr.19: 37537660-37537660 on Build GRCh38
rs776128251 Chr.19: 37537615-37537615 on Build GRCh38
rs768351877 Chr.19: 37537600-37537600 on Build GRCh38
rs896144169 Chr.19: 37537618-37537618 on Build GRCh38
rs867884634 Chr.19: 37537642-37537642 on Build GRCh38
rs886942429 Chr.19: 37537586-37537586 on Build GRCh38
rs370145278 Chr.19: 37537669-37537669 on Build GRCh38
rs745637419 Chr.19: 37537571-37537571 on Build GRCh38
rs759986692 Chr.19: 37537529-37537529 on Build GRCh38
rs766859183 Chr.19: 37537526-37537526 on Build GRCh38
rs778590937 Chr.19: 37537674-37537674 on Build GRCh38
rs758526935 Chr.19: 37537544-37537544 on Build GRCh38
rs755995007 Chr.19: 37537659-37537659 on Build GRCh38
rs764172069 Chr.19: 37537617-37537617 on Build GRCh38
rs777536865 Chr.19: 37537679-37537679 on Build GRCh38
rs757411746 Chr.19: 37537560-37537560 on Build GRCh38
rs867145239 Chr.19: 37537646-37537646 on Build GRCh38
rs761241629 Chr.19: 37537513-37537513 on Build GRCh38

View Full Product Details