Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129162279-129162518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN33
Entrez Gene ID
340348
Gene Name
tetraspanin 33
Gene Aliases
PEN, PEN., TSPAN-33
UniGene
Hs.27267
Gene Chromosome Location
Chr.7: 129144716-129169694 on Build GRCh38
Transcripts
NM_178562.4 XM_006715960.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6548746 c.215G>T p.G72V Chr.7: 129162448-129162448 on Build GRCh38
1673529 c.259C>A p.L87I Chr.7: 129162492-129162492 on Build GRCh38
345903 c.240C>G p.F80L Chr.7: 129162473-129162473 on Build GRCh38
7096748 c.222C>T p.L74L Chr.7: 129162455-129162455 on Build GRCh38
744586 c.264C>A p.R88R Chr.7: 129162497-129162497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762435430 Chr.7: 129162498-129162498 on Build GRCh38
rs769543655 Chr.7: 129162484-129162484 on Build GRCh38
rs768029047 Chr.7: 129162411-129162411 on Build GRCh38
rs762317619 Chr.7: 129162409-129162409 on Build GRCh38
rs367644000 Chr.7: 129162428-129162428 on Build GRCh38
rs764226012 Chr.7: 129162402-129162402 on Build GRCh38
rs374904216 Chr.7: 129162416-129162416 on Build GRCh38
rs750771243 Chr.7: 129162415-129162415 on Build GRCh38
rs768881962 Chr.7: 129162318-129162318 on Build GRCh38
rs548076553 Chr.7: 129162473-129162473 on Build GRCh38
rs775638386 Chr.7: 129162491-129162491 on Build GRCh38
rs756910891 Chr.7: 129162391-129162392 on Build GRCh38
rs1013026196 Chr.7: 129162367-129162367 on Build GRCh38
rs1036667260 Chr.7: 129162413-129162413 on Build GRCh38
rs374105611 Chr.7: 129162379-129162379 on Build GRCh38
rs752391841 Chr.7: 129162426-129162426 on Build GRCh38
rs368039892 Chr.7: 129162380-129162380 on Build GRCh38
rs771048133 Chr.7: 129162400-129162400 on Build GRCh38
rs778750055 Chr.7: 129162421-129162421 on Build GRCh38
rs746924179 Chr.7: 129162455-129162455 on Build GRCh38
rs752060252 Chr.7: 129162405-129162405 on Build GRCh38
rs771582801 Chr.7: 129162345-129162345 on Build GRCh38
rs372845389 Chr.7: 129162479-129162479 on Build GRCh38
rs142963022 Chr.7: 129162497-129162497 on Build GRCh38
rs778592618 Chr.7: 129162344-129162344 on Build GRCh38
rs554484650 Chr.7: 129162325-129162325 on Build GRCh38
rs768626786 Chr.7: 129162496-129162496 on Build GRCh38
rs939001480 Chr.7: 129162478-129162478 on Build GRCh38
rs770815383 Chr.7: 129162458-129162458 on Build GRCh38
rs567610715 Chr.7: 129162364-129162364 on Build GRCh38
rs763219615 Chr.7: 129162495-129162495 on Build GRCh38
rs10464704 Chr.7: 129162322-129162322 on Build GRCh38
rs776716876 Chr.7: 129162441-129162441 on Build GRCh38
rs769954197 Chr.7: 129162382-129162382 on Build GRCh38
rs113176747 Chr.7: 129162348-129162348 on Build GRCh38
rs746954117 Chr.7: 129162366-129162366 on Build GRCh38
rs537857086 Chr.7: 129162384-129162384 on Build GRCh38
rs370368421 Chr.7: 129162397-129162397 on Build GRCh38

View Full Product Details