Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35822667-35822916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF15
Entrez Gene ID
8148
Gene Name
TATA-box binding protein associated factor 15
Gene Aliases
Npl3, RBP56, TAF2N, TAFII68
UniGene
Hs.402752
Gene Chromosome Location
Chr.17: 35809455-35847242 on Build GRCh38
Transcripts
NM_003487.3 NM_139215.2 XM_011525315.1 XM_017025181.1

SNP Detail

SNP ID Chromosome Location
rs771348980 Chr.17: 35822857-35822857 on Build GRCh38
rs912747372 Chr.17: 35822772-35822772 on Build GRCh38
rs755949830 Chr.17: 35822795-35822795 on Build GRCh38
rs376674408 Chr.17: 35822712-35822712 on Build GRCh38
rs148975710 Chr.17: 35822697-35822697 on Build GRCh38
rs762891086 Chr.17: 35822785-35822785 on Build GRCh38
rs771421028 Chr.17: 35822829-35822829 on Build GRCh38
rs770304453 Chr.17: 35822868-35822868 on Build GRCh38
rs78280513 Chr.17: 35822803-35822803 on Build GRCh38
rs140601071 Chr.17: 35822714-35822714 on Build GRCh38
rs868681088 Chr.17: 35822754-35822754 on Build GRCh38
rs777497777 Chr.17: 35822845-35822845 on Build GRCh38
rs776947391 Chr.17: 35822865-35822865 on Build GRCh38
rs779992517 Chr.17: 35822804-35822804 on Build GRCh38
rs777740721 Chr.17: 35822807-35822807 on Build GRCh38
rs755825241 Chr.17: 35822699-35822699 on Build GRCh38
rs369432616 Chr.17: 35822759-35822759 on Build GRCh38
rs766561067 Chr.17: 35822852-35822854 on Build GRCh38
rs765336144 Chr.17: 35822765-35822765 on Build GRCh38
rs771783055 Chr.17: 35822728-35822728 on Build GRCh38
rs777176556 Chr.17: 35822738-35822738 on Build GRCh38
rs986899051 Chr.17: 35822760-35822760 on Build GRCh38
rs772703762 Chr.17: 35822729-35822729 on Build GRCh38
rs780894892 Chr.17: 35822694-35822694 on Build GRCh38
rs371818549 Chr.17: 35822849-35822849 on Build GRCh38
rs750484421 Chr.17: 35822704-35822704 on Build GRCh38
rs369447828 Chr.17: 35822866-35822866 on Build GRCh38
rs747804787 Chr.17: 35822719-35822719 on Build GRCh38
rs752796451 Chr.17: 35822774-35822774 on Build GRCh38
rs775820165 Chr.17: 35822873-35822873 on Build GRCh38
rs989070918 Chr.17: 35822848-35822848 on Build GRCh38
rs764407967 Chr.17: 35822791-35822791 on Build GRCh38
rs140961224 Chr.17: 35822816-35822816 on Build GRCh38
rs751663337 Chr.17: 35822793-35822793 on Build GRCh38
rs763098339 Chr.17: 35822884-35822884 on Build GRCh38
rs754944712 Chr.17: 35822813-35822813 on Build GRCh38
rs759711480 Chr.17: 35822869-35822870 on Build GRCh38
rs962733364 Chr.17: 35822717-35822717 on Build GRCh38
rs751805277 Chr.17: 35822860-35822860 on Build GRCh38
rs780144032 Chr.17: 35822710-35822710 on Build GRCh38
rs954155077 Chr.17: 35822749-35822749 on Build GRCh38
rs756643512 Chr.17: 35822705-35822705 on Build GRCh38
rs761108005 Chr.17: 35822730-35822730 on Build GRCh38
rs749226539 Chr.17: 35822711-35822711 on Build GRCh38
rs533969528 Chr.17: 35822823-35822823 on Build GRCh38
rs182638009 Chr.17: 35822732-35822732 on Build GRCh38

View Full Product Details