Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41388343-41388604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFAF1
Entrez Gene ID
51103
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Gene Aliases
CGI-65, CGI65, CIA30
UniGene
Hs.106529
Gene Chromosome Location
Chr.15: 41387349-41402694 on Build GRCh38
Transcripts
NM_016013.3 NR_045620.1 XM_006720555.2 XM_011521658.1 XM_011521659.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4446654 c.760-3C>T p.? Chr.15: 41388525-41388525 on Build GRCh38
961402 c.790C>T p.R264* Chr.15: 41388492-41388492 on Build GRCh38
699883 c.785C>G p.S262C Chr.15: 41388497-41388497 on Build GRCh38
3769581 c.785C>A p.S262Y Chr.15: 41388497-41388497 on Build GRCh38
6571363 c.780C>T p.F260F Chr.15: 41388502-41388502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138973347 Chr.15: 41388449-41388449 on Build GRCh38
rs139383475 Chr.15: 41388541-41388541 on Build GRCh38
rs770453146 Chr.15: 41388458-41388458 on Build GRCh38
rs766193672 Chr.15: 41388500-41388500 on Build GRCh38
rs771309184 Chr.15: 41388479-41388479 on Build GRCh38
rs781338396 Chr.15: 41388457-41388457 on Build GRCh38
rs200643843 Chr.15: 41388403-41388403 on Build GRCh38
rs995946120 Chr.15: 41388391-41388391 on Build GRCh38
rs755249791 Chr.15: 41388532-41388532 on Build GRCh38
rs767592288 Chr.15: 41388533-41388533 on Build GRCh38
rs765203135 Chr.15: 41388492-41388492 on Build GRCh38
rs372709375 Chr.15: 41388396-41388396 on Build GRCh38
rs1004543092 Chr.15: 41388413-41388413 on Build GRCh38
rs199654703 Chr.15: 41388452-41388452 on Build GRCh38
rs749718737 Chr.15: 41388468-41388468 on Build GRCh38
rs778478444 Chr.15: 41388547-41388547 on Build GRCh38
rs761275085 Chr.15: 41388491-41388491 on Build GRCh38
rs778392729 Chr.15: 41388459-41388459 on Build GRCh38
rs758162413 Chr.15: 41388431-41388431 on Build GRCh38
rs749804636 Chr.15: 41388557-41388557 on Build GRCh38
rs969508672 Chr.15: 41388562-41388562 on Build GRCh38
rs765113176 Chr.15: 41388398-41388398 on Build GRCh38
rs752864871 Chr.15: 41388534-41388534 on Build GRCh38
rs376225447 Chr.15: 41388536-41388536 on Build GRCh38
rs746675778 Chr.15: 41388485-41388485 on Build GRCh38
rs757636199 Chr.15: 41388566-41388566 on Build GRCh38
rs780188968 Chr.15: 41388439-41388439 on Build GRCh38
rs761755490 Chr.15: 41388397-41388397 on Build GRCh38
rs369357784 Chr.15: 41388480-41388480 on Build GRCh38
rs900304150 Chr.15: 41388388-41388388 on Build GRCh38
rs368981046 Chr.15: 41388572-41388572 on Build GRCh38
rs201756363 Chr.15: 41388497-41388497 on Build GRCh38
rs372830971 Chr.15: 41388488-41388488 on Build GRCh38
rs767204079 Chr.15: 41388530-41388530 on Build GRCh38
rs768132279 Chr.15: 41388487-41388487 on Build GRCh38
rs751247707 Chr.15: 41388525-41388525 on Build GRCh38

View Full Product Details