Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107989926-107990181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADH
Entrez Gene ID
3033
Gene Name
hydroxyacyl-CoA dehydrogenase
Gene Aliases
HAD, HADH1, HADHSC, HCDH, HHF4, MSCHAD, SCHAD
UniGene
Hs.438289
Gene Chromosome Location
Chr.4: 107989714-108035175 on Build GRCh38
Transcripts
NM_001184705.2 NM_005327.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420624 c.110T>C p.L37P Chr.4: 107990042-107990042 on Build GRCh38
5958926 c.92delC p.T31fs*7 Chr.4: 107990024-107990024 on Build GRCh38
7144280 c.62C>G p.S21W Chr.4: 107989994-107989994 on Build GRCh38
1209270 c.56C>T p.S19L Chr.4: 107989988-107989988 on Build GRCh38
6420625 c.110T>C p.L37P Chr.4: 107990042-107990042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761180931 Chr.4: 107990059-107990059 on Build GRCh38
rs760216198 Chr.4: 107990087-107990088 on Build GRCh38
rs768651404 Chr.4: 107990158-107990158 on Build GRCh38
rs950643391 Chr.4: 107990083-107990083 on Build GRCh38
rs201600831 Chr.4: 107989982-107989982 on Build GRCh38
rs770646232 Chr.4: 107989969-107989971 on Build GRCh38
rs748128617 Chr.4: 107990008-107990008 on Build GRCh38
rs374248298 Chr.4: 107989979-107989979 on Build GRCh38
rs772843585 Chr.4: 107990070-107990070 on Build GRCh38
rs759364616 Chr.4: 107990089-107990089 on Build GRCh38
rs564000933 Chr.4: 107990153-107990153 on Build GRCh38
rs989646020 Chr.4: 107990139-107990139 on Build GRCh38
rs777924030 Chr.4: 107990024-107990024 on Build GRCh38
rs369131859 Chr.4: 107990132-107990132 on Build GRCh38
rs771772152 Chr.4: 107990066-107990067 on Build GRCh38
rs534966818 Chr.4: 107990071-107990071 on Build GRCh38
rs201094947 Chr.4: 107990092-107990092 on Build GRCh38
rs773991596 Chr.4: 107989982-107989987 on Build GRCh38
rs775923392 Chr.4: 107990056-107990056 on Build GRCh38
rs764746023 Chr.4: 107989973-107989973 on Build GRCh38
rs926097 Chr.4: 107990085-107990085 on Build GRCh38
rs745547559 Chr.4: 107990154-107990154 on Build GRCh38
rs1011049477 Chr.4: 107990138-107990138 on Build GRCh38
rs769224699 Chr.4: 107990060-107990060 on Build GRCh38
rs781319494 Chr.4: 107990004-107990004 on Build GRCh38
rs752625929 Chr.4: 107990108-107990108 on Build GRCh38
rs145521658 Chr.4: 107990035-107990035 on Build GRCh38
rs765688267 Chr.4: 107990106-107990115 on Build GRCh38
rs759111271 Chr.4: 107990065-107990066 on Build GRCh38
rs146017663 Chr.4: 107989950-107989950 on Build GRCh38
rs994692222 Chr.4: 107990038-107990038 on Build GRCh38
rs74428123 Chr.4: 107990031-107990031 on Build GRCh38
rs756215599 Chr.4: 107990009-107990009 on Build GRCh38
rs772452366 Chr.4: 107990048-107990048 on Build GRCh38
rs761268836 Chr.4: 107989967-107989967 on Build GRCh38
rs756127842 Chr.4: 107990110-107990110 on Build GRCh38
rs372068563 Chr.4: 107990105-107990105 on Build GRCh38
rs754944563 Chr.4: 107989995-107989995 on Build GRCh38
rs137853101 Chr.4: 107990050-107990050 on Build GRCh38
rs143346680 Chr.4: 107989970-107989970 on Build GRCh38
rs574138593 Chr.4: 107990157-107990157 on Build GRCh38
rs749406566 Chr.4: 107990027-107990027 on Build GRCh38
rs139920805 Chr.4: 107989966-107989966 on Build GRCh38
rs779135938 Chr.4: 107990032-107990032 on Build GRCh38
rs555570196 Chr.4: 107989984-107989984 on Build GRCh38
rs751280475 Chr.4: 107990087-107990087 on Build GRCh38
rs1003291250 Chr.4: 107989981-107989981 on Build GRCh38
rs957813463 Chr.4: 107990096-107990096 on Build GRCh38
rs886058980 Chr.4: 107989953-107989953 on Build GRCh38
rs900864178 Chr.4: 107990052-107990052 on Build GRCh38
rs767443792 Chr.4: 107990100-107990100 on Build GRCh38
rs879941987 Chr.4: 107990084-107990085 on Build GRCh38
rs749983874 Chr.4: 107989977-107989977 on Build GRCh38
rs915424418 Chr.4: 107990149-107990149 on Build GRCh38

View Full Product Details