Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150676780-150677009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOZ3
Entrez Gene ID
91977
Gene Name
myozenin 3
Gene Aliases
CS-3, CS3, FRP3
UniGene
Hs.91626
Gene Chromosome Location
Chr.5: 150660841-150679368 on Build GRCh38
Transcripts
NM_001122853.2 NM_133371.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5402724 c.714C>T p.A238A Chr.5: 150676833-150676833 on Build GRCh38
4437609 c.685C>T p.R229C Chr.5: 150676804-150676804 on Build GRCh38
7086197 c.684C>G p.L228L Chr.5: 150676803-150676803 on Build GRCh38
4003406 c.702C>T p.F234F Chr.5: 150676821-150676821 on Build GRCh38
6721054 c.667A>G p.S223G Chr.5: 150676786-150676786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146746572 Chr.5: 150676877-150676877 on Build GRCh38
rs769332536 Chr.5: 150676925-150676925 on Build GRCh38
rs748971658 Chr.5: 150676806-150676806 on Build GRCh38
rs755863416 Chr.5: 150676901-150676901 on Build GRCh38
rs372803484 Chr.5: 150676869-150676869 on Build GRCh38
rs769169077 Chr.5: 150676859-150676859 on Build GRCh38
rs189200570 Chr.5: 150676974-150676974 on Build GRCh38
rs201417312 Chr.5: 150676825-150676825 on Build GRCh38
rs552255058 Chr.5: 150676864-150676864 on Build GRCh38
rs745310342 Chr.5: 150676910-150676910 on Build GRCh38
rs201238679 Chr.5: 150676847-150676847 on Build GRCh38
rs747515819 Chr.5: 150676924-150676924 on Build GRCh38
rs752702318 Chr.5: 150676885-150676885 on Build GRCh38
rs538511702 Chr.5: 150676930-150676930 on Build GRCh38
rs976454759 Chr.5: 150676827-150676827 on Build GRCh38
rs756944205 Chr.5: 150676818-150676818 on Build GRCh38
rs922308102 Chr.5: 150676842-150676842 on Build GRCh38
rs144937475 Chr.5: 150676805-150676805 on Build GRCh38
rs767297912 Chr.5: 150676882-150676882 on Build GRCh38
rs763846274 Chr.5: 150676903-150676903 on Build GRCh38
rs1051572178 Chr.5: 150676956-150676956 on Build GRCh38
rs773158786 Chr.5: 150676876-150676876 on Build GRCh38
rs77138054 Chr.5: 150676821-150676821 on Build GRCh38
rs945799178 Chr.5: 150676856-150676856 on Build GRCh38
rs571469572 Chr.5: 150676928-150676928 on Build GRCh38
rs757943119 Chr.5: 150676911-150676911 on Build GRCh38
rs150546422 Chr.5: 150676804-150676804 on Build GRCh38
rs982331924 Chr.5: 150676829-150676829 on Build GRCh38
rs35819533 Chr.5: 150676853-150676854 on Build GRCh38
rs199589261 Chr.5: 150676863-150676863 on Build GRCh38
rs374178828 Chr.5: 150676899-150676899 on Build GRCh38
rs778605607 Chr.5: 150676907-150676907 on Build GRCh38
rs765610710 Chr.5: 150676872-150676872 on Build GRCh38
rs377663921 Chr.5: 150676920-150676920 on Build GRCh38
rs776071245 Chr.5: 150676846-150676846 on Build GRCh38
rs777120044 Chr.5: 150676926-150676926 on Build GRCh38
rs777068489 Chr.5: 150676955-150676955 on Build GRCh38
rs112408743 Chr.5: 150676906-150676906 on Build GRCh38
rs534219526 Chr.5: 150676836-150676836 on Build GRCh38

View Full Product Details