Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5772753-5772990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMP1
Entrez Gene ID
79956
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
FXNA, KIAA1815, bA207C16.3
UniGene
Hs.591078
Gene Chromosome Location
Chr.9: 5764061-5867091 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Gene Symbol
RIC1
Entrez Gene ID
57589
Gene Name
RIC1 homolog, RAB6A GEF complex partner 1
Gene Aliases
CIP150, KIAA1432, bA207C16.1
UniGene
Hs.211520
Gene Chromosome Location
Chr.9: 5629030-5778633 on Build GRCh38
Transcripts
NM_001206557.1 NM_020829.3 XM_005251523.3 XM_011517967.2 XM_011517968.2 XM_017014934.1 XM_017014935.1 XM_017014936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223159 c.3564G>T p.L1188F Chr.9: 5772898-5772898 on Build GRCh38
1109405 c.3641T>C p.I1214T Chr.9: 5772975-5772975 on Build GRCh38
7183688 c.3642C>T p.I1214I Chr.9: 5772976-5772976 on Build GRCh38
1194111 c.3635C>T p.S1212L Chr.9: 5772969-5772969 on Build GRCh38
2777394 c.3604A>G p.I1202V Chr.9: 5772938-5772938 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770223963 Chr.9: 5772955-5772955 on Build GRCh38
rs143909065 Chr.9: 5772846-5772846 on Build GRCh38
rs376399231 Chr.9: 5772904-5772904 on Build GRCh38
rs563306781 Chr.9: 5772837-5772837 on Build GRCh38
rs775211249 Chr.9: 5772859-5772859 on Build GRCh38
rs752907181 Chr.9: 5772924-5772924 on Build GRCh38
rs915532725 Chr.9: 5772927-5772927 on Build GRCh38
rs763608574 Chr.9: 5772861-5772861 on Build GRCh38
rs751634693 Chr.9: 5772936-5772936 on Build GRCh38
rs985213834 Chr.9: 5772797-5772797 on Build GRCh38
rs375660489 Chr.9: 5772850-5772850 on Build GRCh38
rs565348578 Chr.9: 5772941-5772941 on Build GRCh38
rs377503403 Chr.9: 5772790-5772790 on Build GRCh38
rs544986543 Chr.9: 5772811-5772811 on Build GRCh38
rs186010947 Chr.9: 5772931-5772931 on Build GRCh38
rs374069056 Chr.9: 5772852-5772852 on Build GRCh38
rs143563024 Chr.9: 5772906-5772906 on Build GRCh38
rs528246776 Chr.9: 5772866-5772866 on Build GRCh38
rs1027596588 Chr.9: 5772806-5772806 on Build GRCh38
rs62558090 Chr.9: 5772847-5772847 on Build GRCh38
rs950264448 Chr.9: 5772858-5772858 on Build GRCh38
rs925717415 Chr.9: 5772827-5772827 on Build GRCh38
rs760531140 Chr.9: 5772911-5772911 on Build GRCh38
rs780585539 Chr.9: 5772956-5772956 on Build GRCh38
rs146312357 Chr.9: 5772928-5772928 on Build GRCh38
rs1007355586 Chr.9: 5772830-5772830 on Build GRCh38
rs766167027 Chr.9: 5772923-5772923 on Build GRCh38
rs761776572 Chr.9: 5772865-5772865 on Build GRCh38
rs769561996 Chr.9: 5772855-5772855 on Build GRCh38
rs755170468 Chr.9: 5772782-5772782 on Build GRCh38
rs531504574 Chr.9: 5772922-5772922 on Build GRCh38
rs773764228 Chr.9: 5772863-5772863 on Build GRCh38
rs762523121 Chr.9: 5772860-5772860 on Build GRCh38
rs373028666 Chr.9: 5772785-5772785 on Build GRCh38
rs750204830 Chr.9: 5772903-5772903 on Build GRCh38
rs765623332 Chr.9: 5772940-5772940 on Build GRCh38
rs757819865 Chr.9: 5772938-5772938 on Build GRCh38

View Full Product Details