Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119837203-119837464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBPF7
Entrez Gene ID
343505
Gene Name
neuroblastoma breakpoint family member 7
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 119833730-119844880 on Build GRCh38
Transcripts
NM_001047980.2 XM_017001186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521769 c.869G>A p.G290E Chr.1: 119837329-119837329 on Build GRCh38
7093862 c.875C>T p.A292V Chr.1: 119837323-119837323 on Build GRCh38
392478 c.889_890insC p.Q297fs*15 Chr.1: 119837308-119837309 on Build GRCh38
3472287 c.903G>A p.W301* Chr.1: 119837295-119837295 on Build GRCh38
3472288 c.868G>A p.G290R Chr.1: 119837330-119837330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771757530 Chr.1: 119837246-119837246 on Build GRCh38
rs768242788 Chr.1: 119837388-119837388 on Build GRCh38
rs770294697 Chr.1: 119837321-119837321 on Build GRCh38
rs755351632 Chr.1: 119837364-119837364 on Build GRCh38
rs904632920 Chr.1: 119837395-119837395 on Build GRCh38
rs780489813 Chr.1: 119837278-119837278 on Build GRCh38
rs768530126 Chr.1: 119837337-119837337 on Build GRCh38
rs3009184 Chr.1: 119837385-119837385 on Build GRCh38
rs750526242 Chr.1: 119837353-119837353 on Build GRCh38
rs587620126 Chr.1: 119837255-119837255 on Build GRCh38
rs781482694 Chr.1: 119837312-119837312 on Build GRCh38
rs587684153 Chr.1: 119837229-119837229 on Build GRCh38
rs587713645 Chr.1: 119837368-119837368 on Build GRCh38
rs775039259 Chr.1: 119837250-119837250 on Build GRCh38
rs747960561 Chr.1: 119837402-119837402 on Build GRCh38
rs760525007 Chr.1: 119837253-119837253 on Build GRCh38
rs745328106 Chr.1: 119837375-119837375 on Build GRCh38
rs754006822 Chr.1: 119837366-119837366 on Build GRCh38
rs766649702 Chr.1: 119837360-119837360 on Build GRCh38
rs370745985 Chr.1: 119837254-119837254 on Build GRCh38
rs760657752 Chr.1: 119837309-119837309 on Build GRCh38
rs765382026 Chr.1: 119837256-119837256 on Build GRCh38
rs768412597 Chr.1: 119837308-119837309 on Build GRCh38
rs929592803 Chr.1: 119837378-119837378 on Build GRCh38
rs760398333 Chr.1: 119837336-119837336 on Build GRCh38
rs776456555 Chr.1: 119837393-119837393 on Build GRCh38
rs587640132 Chr.1: 119837363-119837363 on Build GRCh38
rs778461293 Chr.1: 119837322-119837322 on Build GRCh38
rs763058548 Chr.1: 119837411-119837411 on Build GRCh38
rs141644184 Chr.1: 119837403-119837403 on Build GRCh38
rs890628792 Chr.1: 119837384-119837384 on Build GRCh38
rs756608687 Chr.1: 119837370-119837370 on Build GRCh38
rs748391368 Chr.1: 119837419-119837419 on Build GRCh38
rs373002211 Chr.1: 119837315-119837315 on Build GRCh38
rs367824497 Chr.1: 119837323-119837323 on Build GRCh38
rs587725648 Chr.1: 119837397-119837397 on Build GRCh38
rs772267088 Chr.1: 119837274-119837274 on Build GRCh38
rs199674818 Chr.1: 119837354-119837354 on Build GRCh38
rs750518568 Chr.1: 119837263-119837263 on Build GRCh38
rs778205589 Chr.1: 119837371-119837371 on Build GRCh38
rs371251675 Chr.1: 119837328-119837328 on Build GRCh38
rs748717842 Chr.1: 119837315-119837315 on Build GRCh38
rs1024175371 Chr.1: 119837233-119837233 on Build GRCh38
rs979883543 Chr.1: 119837279-119837279 on Build GRCh38
rs776385772 Chr.1: 119837339-119837339 on Build GRCh38
rs370417582 Chr.1: 119837252-119837252 on Build GRCh38
rs750267498 Chr.1: 119837227-119837227 on Build GRCh38
rs775140209 Chr.1: 119837334-119837334 on Build GRCh38
rs375400853 Chr.1: 119837343-119837343 on Build GRCh38
rs147914747 Chr.1: 119837381-119837381 on Build GRCh38
rs757911114 Chr.1: 119837380-119837380 on Build GRCh38
rs776480272 Chr.1: 119837376-119837397 on Build GRCh38
rs115348036 Chr.1: 119837309-119837309 on Build GRCh38

View Full Product Details