Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50105475-50105748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD1
Entrez Gene ID
2819
Gene Name
glycerol-3-phosphate dehydrogenase 1
Gene Aliases
GPD-C, GPDH-C, HTGTI
UniGene
Hs.524418
Gene Chromosome Location
Chr.12: 50103819-50111320 on Build GRCh38
Transcripts
NM_001257199.1 NM_005276.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
120911 c.263T>A p.I88N Chr.12: 50105591-50105591 on Build GRCh38
5933047 c.232G>A p.D78N Chr.12: 50105560-50105560 on Build GRCh38
4042583 c.242A>G p.Q81R Chr.12: 50105570-50105570 on Build GRCh38
1188492 c.253G>A p.D85N Chr.12: 50105581-50105581 on Build GRCh38
4967998 c.319G>T p.G107C Chr.12: 50105647-50105647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763610835 Chr.12: 50105707-50105707 on Build GRCh38
rs779973836 Chr.12: 50105726-50105726 on Build GRCh38
rs374426834 Chr.12: 50105620-50105620 on Build GRCh38
rs141658302 Chr.12: 50105590-50105590 on Build GRCh38
rs200773155 Chr.12: 50105535-50105535 on Build GRCh38
rs771458779 Chr.12: 50105658-50105658 on Build GRCh38
rs147914698 Chr.12: 50105564-50105564 on Build GRCh38
rs375009098 Chr.12: 50105725-50105725 on Build GRCh38
rs777046660 Chr.12: 50105563-50105565 on Build GRCh38
rs761496413 Chr.12: 50105716-50105716 on Build GRCh38
rs758135465 Chr.12: 50105593-50105593 on Build GRCh38
rs767351255 Chr.12: 50105722-50105722 on Build GRCh38
rs11541968 Chr.12: 50105572-50105572 on Build GRCh38
rs531712569 Chr.12: 50105523-50105523 on Build GRCh38
rs1023426385 Chr.12: 50105682-50105682 on Build GRCh38
rs747209928 Chr.12: 50105652-50105652 on Build GRCh38
rs776072071 Chr.12: 50105677-50105677 on Build GRCh38
rs773695657 Chr.12: 50105560-50105560 on Build GRCh38
rs781120676 Chr.12: 50105540-50105540 on Build GRCh38
rs1024035022 Chr.12: 50105674-50105674 on Build GRCh38
rs773088027 Chr.12: 50105578-50105578 on Build GRCh38
rs768754769 Chr.12: 50105547-50105547 on Build GRCh38
rs757008468 Chr.12: 50105539-50105539 on Build GRCh38
rs887175208 Chr.12: 50105566-50105566 on Build GRCh38
rs758434100 Chr.12: 50105729-50105729 on Build GRCh38
rs17848998 Chr.12: 50105628-50105628 on Build GRCh38
rs772126813 Chr.12: 50105643-50105643 on Build GRCh38
rs571123918 Chr.12: 50105667-50105667 on Build GRCh38
rs756025107 Chr.12: 50105623-50105623 on Build GRCh38
rs200622182 Chr.12: 50105696-50105696 on Build GRCh38
rs777110187 Chr.12: 50105661-50105661 on Build GRCh38
rs751202719 Chr.12: 50105714-50105714 on Build GRCh38
rs753755182 Chr.12: 50105727-50105727 on Build GRCh38
rs995015035 Chr.12: 50105526-50105526 on Build GRCh38
rs752433970 Chr.12: 50105527-50105527 on Build GRCh38
rs34537997 Chr.12: 50105543-50105544 on Build GRCh38
rs367611175 Chr.12: 50105534-50105534 on Build GRCh38
rs1013854308 Chr.12: 50105609-50105609 on Build GRCh38
rs745757014 Chr.12: 50105544-50105544 on Build GRCh38
rs777679621 Chr.12: 50105530-50105530 on Build GRCh38
rs201717429 Chr.12: 50105502-50105502 on Build GRCh38
rs754553503 Chr.12: 50105524-50105524 on Build GRCh38
rs772173280 Chr.12: 50105558-50105558 on Build GRCh38
rs192019432 Chr.12: 50105619-50105619 on Build GRCh38
rs763657369 Chr.12: 50105595-50105595 on Build GRCh38
rs748419979 Chr.12: 50105548-50105548 on Build GRCh38
rs764795976 Chr.12: 50105525-50105525 on Build GRCh38
rs199633670 Chr.12: 50105640-50105640 on Build GRCh38
rs757025524 Chr.12: 50105616-50105616 on Build GRCh38
rs766712596 Chr.12: 50105573-50105573 on Build GRCh38
rs757963092 Chr.12: 50105528-50105528 on Build GRCh38
rs970211430 Chr.12: 50105702-50105702 on Build GRCh38
rs762382065 Chr.12: 50105706-50105706 on Build GRCh38
rs773324714 Chr.12: 50105649-50105649 on Build GRCh38
rs745846984 Chr.12: 50105622-50105622 on Build GRCh38
rs1128867 Chr.12: 50105665-50105665 on Build GRCh38
rs752239622 Chr.12: 50105592-50105592 on Build GRCh38
rs537531194 Chr.12: 50105724-50105724 on Build GRCh38
rs534115061 Chr.12: 50105608-50105608 on Build GRCh38
rs761306104 Chr.12: 50105515-50105515 on Build GRCh38
rs767967933 Chr.12: 50105498-50105498 on Build GRCh38
rs760176358 Chr.12: 50105664-50105664 on Build GRCh38
rs760086348 Chr.12: 50105580-50105580 on Build GRCh38

View Full Product Details