Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124012248-124012473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX2
Entrez Gene ID
9355
Gene Name
LIM homeobox 2
Gene Aliases
LH2, hLhx2
UniGene
Hs.696425
Gene Chromosome Location
Chr.9: 124011610-124033163 on Build GRCh38
Transcripts
NM_004789.3 XM_006717323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6550465 c.12C>T p.H4H Chr.9: 124012360-124012360 on Build GRCh38
6352903 c.49G>C p.E17Q Chr.9: 124012397-124012397 on Build GRCh38
4697912 c.57C>T p.D19D Chr.9: 124012405-124012405 on Build GRCh38
6412884 c.76G>T p.A26S Chr.9: 124012424-124012424 on Build GRCh38
1105035 c.73G>A p.E25K Chr.9: 124012421-124012421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775952839 Chr.9: 124012332-124012332 on Build GRCh38
rs541906506 Chr.9: 124012270-124012270 on Build GRCh38
rs905603997 Chr.9: 124012429-124012429 on Build GRCh38
rs750239855 Chr.9: 124012296-124012296 on Build GRCh38
rs879184022 Chr.9: 124012326-124012326 on Build GRCh38
rs760154535 Chr.9: 124012325-124012325 on Build GRCh38
rs923570861 Chr.9: 124012284-124012284 on Build GRCh38
rs563485024 Chr.9: 124012306-124012306 on Build GRCh38
rs752033219 Chr.9: 124012372-124012372 on Build GRCh38
rs891476244 Chr.9: 124012369-124012369 on Build GRCh38
rs1001401446 Chr.9: 124012456-124012456 on Build GRCh38
rs968124826 Chr.9: 124012394-124012394 on Build GRCh38
rs767900210 Chr.9: 124012401-124012401 on Build GRCh38
rs754959160 Chr.9: 124012435-124012435 on Build GRCh38
rs967525166 Chr.9: 124012329-124012329 on Build GRCh38
rs751231966 Chr.9: 124012330-124012331 on Build GRCh38
rs752934468 Chr.9: 124012403-124012403 on Build GRCh38
rs1039603816 Chr.9: 124012275-124012275 on Build GRCh38
rs924111235 Chr.9: 124012417-124012417 on Build GRCh38
rs960676643 Chr.9: 124012292-124012292 on Build GRCh38
rs756249258 Chr.9: 124012412-124012412 on Build GRCh38
rs763527047 Chr.9: 124012342-124012342 on Build GRCh38
rs755380207 Chr.9: 124012397-124012397 on Build GRCh38
rs530813615 Chr.9: 124012344-124012344 on Build GRCh38
rs763631459 Chr.9: 124012327-124012327 on Build GRCh38
rs980010337 Chr.9: 124012411-124012411 on Build GRCh38
rs1042853209 Chr.9: 124012425-124012425 on Build GRCh38

View Full Product Details