Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16852562-16852823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP7
Entrez Gene ID
5931
Gene Name
RB binding protein 7, chromatin remodeling factor
Gene Aliases
RbAp46
UniGene
Hs.495755
Gene Chromosome Location
Chr.X: 16844650-16870411 on Build GRCh38
Transcripts
NM_001198719.1 NM_002893.3

SNP Detail

SNP ID Chromosome Location
rs749468933 Chr.X: 16852643-16852643 on Build GRCh38
rs778991028 Chr.X: 16852654-16852654 on Build GRCh38
rs762369480 Chr.X: 16852732-16852732 on Build GRCh38
rs759738532 Chr.X: 16852681-16852681 on Build GRCh38
rs764812881 Chr.X: 16852715-16852715 on Build GRCh38
rs368343853 Chr.X: 16852791-16852791 on Build GRCh38
rs766098154 Chr.X: 16852652-16852652 on Build GRCh38
rs770994288 Chr.X: 16852640-16852640 on Build GRCh38
rs538515609 Chr.X: 16852694-16852694 on Build GRCh38
rs180940768 Chr.X: 16852764-16852764 on Build GRCh38
rs755992987 Chr.X: 16852761-16852761 on Build GRCh38
rs761465578 Chr.X: 16852709-16852709 on Build GRCh38
rs370817118 Chr.X: 16852701-16852701 on Build GRCh38
rs868486471 Chr.X: 16852762-16852762 on Build GRCh38
rs774309971 Chr.X: 16852673-16852673 on Build GRCh38
rs200419169 Chr.X: 16852755-16852755 on Build GRCh38
rs187154873 Chr.X: 16852797-16852797 on Build GRCh38
rs780895806 Chr.X: 16852744-16852744 on Build GRCh38
rs769095277 Chr.X: 16852704-16852704 on Build GRCh38
rs368553513 Chr.X: 16852674-16852674 on Build GRCh38
rs777169033 Chr.X: 16852708-16852708 on Build GRCh38
rs189058844 Chr.X: 16852626-16852626 on Build GRCh38
rs771061288 Chr.X: 16852645-16852645 on Build GRCh38
rs926891183 Chr.X: 16852672-16852672 on Build GRCh38
rs750152508 Chr.X: 16852724-16852724 on Build GRCh38
rs746142082 Chr.X: 16852782-16852782 on Build GRCh38
rs762793373 Chr.X: 16852619-16852619 on Build GRCh38
rs60555681 Chr.X: 16852743-16852743 on Build GRCh38
rs1012562103 Chr.X: 16852684-16852684 on Build GRCh38
rs765919525 Chr.X: 16852734-16852734 on Build GRCh38
rs776110763 Chr.X: 16852700-16852700 on Build GRCh38
rs767000899 Chr.X: 16852784-16852784 on Build GRCh38

View Full Product Details