Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23007606-23007863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOBTB2
Entrez Gene ID
23221
Gene Name
Rho related BTB domain containing 2
Gene Aliases
DBC2
UniGene
Hs.372688
Gene Chromosome Location
Chr.8: 22987406-23020197 on Build GRCh38
Transcripts
NM_001160036.1 NM_001160037.1 NM_015178.2 XM_017013251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1098185 c.1381C>T p.R461C Chr.8: 23007626-23007626 on Build GRCh38
6874802 c.1368G>A p.E456E Chr.8: 23007613-23007613 on Build GRCh38
136652 c.1366G>A p.E456K Chr.8: 23007611-23007611 on Build GRCh38
2786474 c.1450G>A p.V484I Chr.8: 23007695-23007695 on Build GRCh38
4527123 c.1375G>C p.D459H Chr.8: 23007620-23007620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757772975 Chr.8: 23007713-23007713 on Build GRCh38
rs376143315 Chr.8: 23007770-23007770 on Build GRCh38
rs774442722 Chr.8: 23007782-23007782 on Build GRCh38
rs777766985 Chr.8: 23007695-23007695 on Build GRCh38
rs995670907 Chr.8: 23007743-23007743 on Build GRCh38
rs984599000 Chr.8: 23007768-23007768 on Build GRCh38
rs776228984 Chr.8: 23007751-23007751 on Build GRCh38
rs769323162 Chr.8: 23007765-23007765 on Build GRCh38
rs373166557 Chr.8: 23007766-23007766 on Build GRCh38
rs779401316 Chr.8: 23007734-23007734 on Build GRCh38
rs774235840 Chr.8: 23007758-23007758 on Build GRCh38
rs1022988354 Chr.8: 23007646-23007646 on Build GRCh38
rs752989589 Chr.8: 23007654-23007654 on Build GRCh38
rs1036684641 Chr.8: 23007803-23007803 on Build GRCh38
rs2430815 Chr.8: 23007807-23007807 on Build GRCh38
rs960359874 Chr.8: 23007730-23007730 on Build GRCh38
rs550141841 Chr.8: 23007735-23007735 on Build GRCh38
rs968291983 Chr.8: 23007660-23007660 on Build GRCh38
rs975771686 Chr.8: 23007821-23007821 on Build GRCh38
rs759354241 Chr.8: 23007796-23007796 on Build GRCh38
rs918608907 Chr.8: 23007805-23007805 on Build GRCh38
rs969989376 Chr.8: 23007706-23007706 on Build GRCh38
rs749789347 Chr.8: 23007709-23007709 on Build GRCh38
rs369972623 Chr.8: 23007783-23007783 on Build GRCh38
rs150932568 Chr.8: 23007694-23007694 on Build GRCh38
rs142549794 Chr.8: 23007643-23007643 on Build GRCh38
rs912525185 Chr.8: 23007806-23007806 on Build GRCh38
rs370244536 Chr.8: 23007771-23007771 on Build GRCh38
rs1027503610 Chr.8: 23007750-23007750 on Build GRCh38
rs544481877 Chr.8: 23007781-23007781 on Build GRCh38
rs767789125 Chr.8: 23007653-23007653 on Build GRCh38
rs771192574 Chr.8: 23007680-23007681 on Build GRCh38

View Full Product Details