Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37612684-37612957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPO1
Entrez Gene ID
284654
Gene Name
R-spondin 1
Gene Aliases
CRISTIN3, RSPO
UniGene
Hs.135015
Gene Chromosome Location
Chr.1: 37611350-37634923 on Build GRCh38
Transcripts
NM_001038633.3 NM_001242908.1 NM_001242909.1 NM_001242910.1 XM_006710583.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7388283 c.719G>A p.R240Q Chr.1: 37612828-37612828 on Build GRCh38
426214 c.654C>A p.G218G Chr.1: 37612893-37612893 on Build GRCh38
1720304 c.750_752delGCA p.Q251delQ Chr.1: 37612795-37612797 on Build GRCh38
5538410 c.633G>A p.K211K Chr.1: 37612914-37612914 on Build GRCh38
1224344 c.626G>T p.G209V Chr.1: 37612921-37612921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568772955 Chr.1: 37612749-37612749 on Build GRCh38
rs201963347 Chr.1: 37612843-37612843 on Build GRCh38
rs756221900 Chr.1: 37612747-37612747 on Build GRCh38
rs776626302 Chr.1: 37612918-37612918 on Build GRCh38
rs776514324 Chr.1: 37612722-37612722 on Build GRCh38
rs779652827 Chr.1: 37612774-37612774 on Build GRCh38
rs775859628 Chr.1: 37612902-37612902 on Build GRCh38
rs746232403 Chr.1: 37612765-37612765 on Build GRCh38
rs372057847 Chr.1: 37612891-37612891 on Build GRCh38
rs763326013 Chr.1: 37612932-37612932 on Build GRCh38
rs375231071 Chr.1: 37612908-37612908 on Build GRCh38
rs202089760 Chr.1: 37612861-37612861 on Build GRCh38
rs772927728 Chr.1: 37612823-37612823 on Build GRCh38
rs546859528 Chr.1: 37612731-37612731 on Build GRCh38
rs767823808 Chr.1: 37612736-37612736 on Build GRCh38
rs760850808 Chr.1: 37612838-37612838 on Build GRCh38
rs1014157474 Chr.1: 37612723-37612723 on Build GRCh38
rs182482113 Chr.1: 37612889-37612889 on Build GRCh38
rs758725998 Chr.1: 37612858-37612858 on Build GRCh38
rs757646904 Chr.1: 37612753-37612753 on Build GRCh38
rs573351631 Chr.1: 37612895-37612895 on Build GRCh38
rs754242025 Chr.1: 37612752-37612752 on Build GRCh38
rs267598586 Chr.1: 37612881-37612881 on Build GRCh38
rs749116749 Chr.1: 37612892-37612892 on Build GRCh38
rs534011478 Chr.1: 37612921-37612921 on Build GRCh38
rs750795562 Chr.1: 37612853-37612853 on Build GRCh38
rs769753826 Chr.1: 37612822-37612822 on Build GRCh38
rs370389098 Chr.1: 37612904-37612904 on Build GRCh38
rs759731020 Chr.1: 37612735-37612735 on Build GRCh38
rs376406082 Chr.1: 37612815-37612815 on Build GRCh38
rs779213780 Chr.1: 37612755-37612755 on Build GRCh38
rs866646848 Chr.1: 37612903-37612903 on Build GRCh38
rs751158921 Chr.1: 37612871-37612871 on Build GRCh38
rs775804545 Chr.1: 37612836-37612836 on Build GRCh38
rs563949840 Chr.1: 37612794-37612795 on Build GRCh38
rs750897150 Chr.1: 37612931-37612931 on Build GRCh38
rs780621142 Chr.1: 37612888-37612888 on Build GRCh38
rs1040571358 Chr.1: 37612876-37612876 on Build GRCh38
rs561199140 Chr.1: 37612734-37612734 on Build GRCh38
rs775992037 Chr.1: 37612899-37612900 on Build GRCh38
rs769657393 Chr.1: 37612890-37612890 on Build GRCh38
rs765754433 Chr.1: 37612851-37612851 on Build GRCh38
rs764203076 Chr.1: 37612842-37612842 on Build GRCh38
rs746502792 Chr.1: 37612712-37612712 on Build GRCh38
rs762285891 Chr.1: 37612848-37612848 on Build GRCh38
rs749525234 Chr.1: 37612802-37612802 on Build GRCh38
rs746335777 Chr.1: 37612737-37612771 on Build GRCh38
rs865863233 Chr.1: 37612849-37612849 on Build GRCh38
rs761694625 Chr.1: 37612725-37612725 on Build GRCh38
rs535890035 Chr.1: 37612766-37612766 on Build GRCh38
rs376389761 Chr.1: 37612893-37612893 on Build GRCh38
rs997410528 Chr.1: 37612831-37612831 on Build GRCh38
rs768110214 Chr.1: 37612803-37612803 on Build GRCh38
rs754617519 Chr.1: 37612886-37612886 on Build GRCh38
rs765697741 Chr.1: 37612922-37612922 on Build GRCh38
rs764869923 Chr.1: 37612729-37612729 on Build GRCh38
rs967203904 Chr.1: 37612744-37612744 on Build GRCh38
rs768623621 Chr.1: 37612713-37612713 on Build GRCh38
rs200509081 Chr.1: 37612821-37612821 on Build GRCh38
rs372532240 Chr.1: 37612829-37612829 on Build GRCh38
rs752862829 Chr.1: 37612741-37612741 on Build GRCh38
rs61748625 Chr.1: 37612828-37612828 on Build GRCh38

View Full Product Details