Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92303930-92304185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
UniGene
Hs.49105
Gene Chromosome Location
Chr.1: 92245786-92441936 on Build GRCh38
Transcripts
XM_011540546.2 NM_024813.2 XM_005271223.2 XM_011542167.2 XM_017002362.1 XM_017002363.1 XM_017002364.1

Gene Symbol
RPAP2
Entrez Gene ID
79871
Gene Name
RNA polymerase II associated protein 2
Gene Aliases
C1orf82, Rtr1
UniGene
Hs.444421
Gene Chromosome Location
Chr.1: 92298965-92388175 on Build GRCh38
Transcripts
XM_011540546.2 NM_024813.2 XM_005271223.2 XM_011542167.2 XM_017002362.1 XM_017002363.1 XM_017002364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6434721 c.326T>C p.L109P Chr.1: 92304068-92304068 on Build GRCh38
1748664 c.256C>T p.H86Y Chr.1: 92303998-92303998 on Build GRCh38
4722515 c.297C>A p.L99L Chr.1: 92304039-92304039 on Build GRCh38
7388089 c.235-1G>T p.? Chr.1: 92303976-92303976 on Build GRCh38
4411831 c.295C>A p.L99I Chr.1: 92304037-92304037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764038548 Chr.1: 92304087-92304088 on Build GRCh38
rs774278468 Chr.1: 92304013-92304013 on Build GRCh38
rs753014609 Chr.1: 92304025-92304025 on Build GRCh38
rs531745273 Chr.1: 92304049-92304049 on Build GRCh38
rs768458600 Chr.1: 92304109-92304109 on Build GRCh38
rs1055653640 Chr.1: 92304037-92304038 on Build GRCh38
rs776290519 Chr.1: 92303983-92303983 on Build GRCh38
rs992982028 Chr.1: 92304076-92304076 on Build GRCh38
rs757687131 Chr.1: 92304083-92304083 on Build GRCh38
rs775332656 Chr.1: 92304003-92304003 on Build GRCh38
rs749041926 Chr.1: 92304106-92304106 on Build GRCh38
rs770430332 Chr.1: 92303969-92303969 on Build GRCh38
rs775066670 Chr.1: 92304098-92304098 on Build GRCh38
rs746502898 Chr.1: 92303966-92303966 on Build GRCh38
rs374105804 Chr.1: 92304023-92304023 on Build GRCh38
rs753749410 Chr.1: 92304115-92304115 on Build GRCh38
rs755316739 Chr.1: 92304022-92304022 on Build GRCh38
rs925951850 Chr.1: 92304137-92304137 on Build GRCh38
rs760346445 Chr.1: 92304050-92304051 on Build GRCh38
rs926364973 Chr.1: 92304035-92304035 on Build GRCh38
rs77762919 Chr.1: 92304012-92304012 on Build GRCh38
rs771566224 Chr.1: 92304008-92304009 on Build GRCh38
rs574533018 Chr.1: 92304004-92304004 on Build GRCh38
rs1030376104 Chr.1: 92304152-92304152 on Build GRCh38
rs775357856 Chr.1: 92304040-92304041 on Build GRCh38
rs761849959 Chr.1: 92304127-92304127 on Build GRCh38
rs769408465 Chr.1: 92303992-92303992 on Build GRCh38
rs761315297 Chr.1: 92304028-92304028 on Build GRCh38
rs563216903 Chr.1: 92304095-92304095 on Build GRCh38
rs780724140 Chr.1: 92304089-92304089 on Build GRCh38
rs778170607 Chr.1: 92304033-92304033 on Build GRCh38
rs184276869 Chr.1: 92304096-92304096 on Build GRCh38
rs753937033 Chr.1: 92304016-92304016 on Build GRCh38
rs914069721 Chr.1: 92303964-92303964 on Build GRCh38
rs745572851 Chr.1: 92303988-92303988 on Build GRCh38
rs545349551 Chr.1: 92304153-92304153 on Build GRCh38
rs897648399 Chr.1: 92304124-92304124 on Build GRCh38
rs773995248 Chr.1: 92304120-92304120 on Build GRCh38

View Full Product Details