Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76625333-76625576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GALNAC1
Entrez Gene ID
55808
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
HSY11339, SIAT7A, ST6GalNAcI, STYI
UniGene
Hs.105352
Gene Chromosome Location
Chr.17: 76617769-76643838 on Build GRCh38
Transcripts
NM_001289107.1 NM_018414.4 NR_110309.1 XM_011524995.2 XM_011524996.2 XM_017024842.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522588 c.1698C>T p.N566N Chr.17: 76625435-76625435 on Build GRCh38
4835316 c.1728G>A p.W576* Chr.17: 76625405-76625405 on Build GRCh38
984740 c.1672T>C p.W558R Chr.17: 76625461-76625461 on Build GRCh38
3522587 c.1741G>A p.D581N Chr.17: 76625392-76625392 on Build GRCh38
984738 c.1758G>A p.R586R Chr.17: 76625375-76625375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373474836 Chr.17: 76625424-76625424 on Build GRCh38
rs780932173 Chr.17: 76625510-76625510 on Build GRCh38
rs1021413544 Chr.17: 76625398-76625398 on Build GRCh38
rs755142819 Chr.17: 76625353-76625353 on Build GRCh38
rs559542547 Chr.17: 76625490-76625490 on Build GRCh38
rs201887138 Chr.17: 76625354-76625354 on Build GRCh38
rs149996373 Chr.17: 76625393-76625393 on Build GRCh38
rs112544141 Chr.17: 76625542-76625542 on Build GRCh38
rs371455150 Chr.17: 76625376-76625376 on Build GRCh38
rs200189269 Chr.17: 76625526-76625526 on Build GRCh38
rs778767132 Chr.17: 76625475-76625475 on Build GRCh38
rs779676226 Chr.17: 76625388-76625388 on Build GRCh38
rs777426595 Chr.17: 76625370-76625370 on Build GRCh38
rs545290323 Chr.17: 76625451-76625451 on Build GRCh38
rs566898658 Chr.17: 76625501-76625501 on Build GRCh38
rs867272064 Chr.17: 76625401-76625401 on Build GRCh38
rs771434024 Chr.17: 76625522-76625522 on Build GRCh38
rs886366142 Chr.17: 76625416-76625416 on Build GRCh38
rs200682151 Chr.17: 76625456-76625456 on Build GRCh38
rs569310023 Chr.17: 76625552-76625552 on Build GRCh38
rs764422256 Chr.17: 76625455-76625455 on Build GRCh38
rs771586239 Chr.17: 76625384-76625384 on Build GRCh38
rs150577421 Chr.17: 76625365-76625365 on Build GRCh38
rs144687878 Chr.17: 76625527-76625527 on Build GRCh38
rs369878924 Chr.17: 76625516-76625516 on Build GRCh38
rs140855041 Chr.17: 76625523-76625523 on Build GRCh38
rs369261157 Chr.17: 76625408-76625408 on Build GRCh38
rs752875565 Chr.17: 76625364-76625364 on Build GRCh38
rs145873819 Chr.17: 76625377-76625377 on Build GRCh38
rs771526332 Chr.17: 76625417-76625417 on Build GRCh38
rs760830792 Chr.17: 76625539-76625539 on Build GRCh38
rs746582013 Chr.17: 76625505-76625505 on Build GRCh38
rs753669733 Chr.17: 76625372-76625372 on Build GRCh38
rs753578689 Chr.17: 76625461-76625461 on Build GRCh38
rs767481502 Chr.17: 76625433-76625433 on Build GRCh38
rs375478556 Chr.17: 76625492-76625492 on Build GRCh38
rs148549229 Chr.17: 76625491-76625491 on Build GRCh38
rs144223429 Chr.17: 76625425-76625425 on Build GRCh38
rs757073017 Chr.17: 76625465-76625465 on Build GRCh38
rs775409453 Chr.17: 76625537-76625537 on Build GRCh38
rs2286595 Chr.17: 76625553-76625553 on Build GRCh38
rs183186412 Chr.17: 76625400-76625400 on Build GRCh38
rs768148497 Chr.17: 76625509-76625509 on Build GRCh38
rs373184306 Chr.17: 76625536-76625536 on Build GRCh38
rs746641827 Chr.17: 76625392-76625392 on Build GRCh38
rs1056255219 Chr.17: 76625422-76625422 on Build GRCh38
rs770048230 Chr.17: 76625436-76625436 on Build GRCh38
rs756335445 Chr.17: 76625454-76625454 on Build GRCh38
rs1029341862 Chr.17: 76625426-76625426 on Build GRCh38
rs745319479 Chr.17: 76625379-76625379 on Build GRCh38

View Full Product Details