Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 97736230-97736501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAC1
Entrez Gene ID
6863
Gene Name
tachykinin precursor 1
Gene Aliases
Hs.2563, NK2, NKNA, NPK, TAC2
UniGene
Hs.2563
Gene Chromosome Location
Chr.7: 97731959-97740472 on Build GRCh38
Transcripts
NM_003182.2 NM_013996.2 NM_013997.2 NM_013998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3642622 c.322G>A p.G108S Chr.7: 97736331-97736331 on Build GRCh38
6178932 c.332C>G p.A111G Chr.7: 97736341-97736341 on Build GRCh38
5062693 c.290G>T p.R97I Chr.7: 97736299-97736299 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997942933 Chr.7: 97736439-97736439 on Build GRCh38
rs762430556 Chr.7: 97736376-97736376 on Build GRCh38
rs899766346 Chr.7: 97736406-97736406 on Build GRCh38
rs778794091 Chr.7: 97736290-97736290 on Build GRCh38
rs769124233 Chr.7: 97736338-97736338 on Build GRCh38
rs767160685 Chr.7: 97736392-97736392 on Build GRCh38
rs746121957 Chr.7: 97736292-97736292 on Build GRCh38
rs777204544 Chr.7: 97736352-97736352 on Build GRCh38
rs200093351 Chr.7: 97736373-97736373 on Build GRCh38
rs191687379 Chr.7: 97736455-97736455 on Build GRCh38
rs756387093 Chr.7: 97736282-97736282 on Build GRCh38
rs540282329 Chr.7: 97736305-97736305 on Build GRCh38
rs1029462706 Chr.7: 97736463-97736463 on Build GRCh38
rs754740388 Chr.7: 97736263-97736263 on Build GRCh38
rs747444005 Chr.7: 97736331-97736331 on Build GRCh38
rs751212163 Chr.7: 97736381-97736381 on Build GRCh38
rs142031557 Chr.7: 97736303-97736303 on Build GRCh38
rs780391486 Chr.7: 97736316-97736316 on Build GRCh38
rs371024959 Chr.7: 97736379-97736379 on Build GRCh38
rs186329170 Chr.7: 97736363-97736363 on Build GRCh38
rs757372729 Chr.7: 97736282-97736282 on Build GRCh38
rs372377656 Chr.7: 97736264-97736264 on Build GRCh38
rs946806371 Chr.7: 97736374-97736374 on Build GRCh38
rs539105664 Chr.7: 97736379-97736382 on Build GRCh38
rs746181537 Chr.7: 97736459-97736459 on Build GRCh38

View Full Product Details