Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160010632-160010897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC1
Entrez Gene ID
7265
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
TPR1
UniGene
Hs.519718
Gene Chromosome Location
Chr.5: 160009100-160065545 on Build GRCh38
Transcripts
NM_001282500.1 NM_003314.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696669 c.277G>A p.D93N Chr.5: 160010805-160010805 on Build GRCh38
5402931 c.324G>C p.E108D Chr.5: 160010852-160010852 on Build GRCh38
5617274 c.300G>A p.L100L Chr.5: 160010828-160010828 on Build GRCh38
242667 c.212A>T p.E71V Chr.5: 160010740-160010740 on Build GRCh38
5744868 c.234G>T p.K78N Chr.5: 160010762-160010762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559624090 Chr.5: 160010678-160010678 on Build GRCh38
rs769425474 Chr.5: 160010817-160010817 on Build GRCh38
rs768210255 Chr.5: 160010711-160010711 on Build GRCh38
rs149889700 Chr.5: 160010783-160010783 on Build GRCh38
rs752108060 Chr.5: 160010794-160010794 on Build GRCh38
rs557280478 Chr.5: 160010727-160010727 on Build GRCh38
rs774876013 Chr.5: 160010706-160010708 on Build GRCh38
rs773006026 Chr.5: 160010742-160010742 on Build GRCh38
rs754164804 Chr.5: 160010670-160010670 on Build GRCh38
rs147640958 Chr.5: 160010828-160010828 on Build GRCh38
rs762024262 Chr.5: 160010829-160010830 on Build GRCh38
rs756332105 Chr.5: 160010659-160010659 on Build GRCh38
rs200775911 Chr.5: 160010732-160010732 on Build GRCh38
rs150711044 Chr.5: 160010699-160010699 on Build GRCh38
rs1038538403 Chr.5: 160010837-160010837 on Build GRCh38
rs754449659 Chr.5: 160010797-160010797 on Build GRCh38
rs141210936 Chr.5: 160010682-160010682 on Build GRCh38
rs774401704 Chr.5: 160010870-160010870 on Build GRCh38
rs774489584 Chr.5: 160010762-160010762 on Build GRCh38
rs760818093 Chr.5: 160010763-160010763 on Build GRCh38
rs780522096 Chr.5: 160010787-160010787 on Build GRCh38
rs769358691 Chr.5: 160010734-160010734 on Build GRCh38
rs757531820 Chr.5: 160010672-160010672 on Build GRCh38
rs757450483 Chr.5: 160010771-160010771 on Build GRCh38
rs41275307 Chr.5: 160010755-160010755 on Build GRCh38
rs749099073 Chr.5: 160010832-160010832 on Build GRCh38
rs754003511 Chr.5: 160010769-160010769 on Build GRCh38
rs774572353 Chr.5: 160010806-160010806 on Build GRCh38
rs770682888 Chr.5: 160010840-160010840 on Build GRCh38
rs17849377 Chr.5: 160010829-160010829 on Build GRCh38
rs984157514 Chr.5: 160010846-160010846 on Build GRCh38
rs777965550 Chr.5: 160010747-160010747 on Build GRCh38
rs773655875 Chr.5: 160010859-160010860 on Build GRCh38
rs764309520 Chr.5: 160010766-160010766 on Build GRCh38
rs758754532 Chr.5: 160010784-160010784 on Build GRCh38
rs556412884 Chr.5: 160010706-160010706 on Build GRCh38
rs139856144 Chr.5: 160010674-160010674 on Build GRCh38
rs765391613 Chr.5: 160010772-160010772 on Build GRCh38
rs11555804 Chr.5: 160010760-160010760 on Build GRCh38
rs768072394 Chr.5: 160010859-160010862 on Build GRCh38
rs752760728 Chr.5: 160010656-160010656 on Build GRCh38
rs747627523 Chr.5: 160010815-160010815 on Build GRCh38
rs1020600997 Chr.5: 160010675-160010675 on Build GRCh38
rs532228909 Chr.5: 160010666-160010666 on Build GRCh38
rs964709828 Chr.5: 160010667-160010667 on Build GRCh38
rs758882425 Chr.5: 160010683-160010683 on Build GRCh38

View Full Product Details