Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46494622-46494857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNMAL2
Entrez Gene ID
57469
Gene Name
paraneoplastic Ma antigen family-like 2
Gene Aliases
-
UniGene
Hs.7193
Gene Chromosome Location
Chr.19: 46491191-46495912 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Gene Symbol
PPP5D1
Entrez Gene ID
100506012
Gene Name
PPP5 tetratricopeptide repeat domain containing 1
Gene Aliases
-
UniGene
Hs.671661
Gene Chromosome Location
Chr.19: 46480788-46601200 on Build GRCh38
Transcripts
NM_020709.2 NM_001205281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998439 c.160G>A p.A54T Chr.19: 46494792-46494792 on Build GRCh38
7285300 c.676A>G p.T226A Chr.19: 46494790-46494790 on Build GRCh38
5744193 c.677C>T p.T226M Chr.19: 46494789-46494789 on Build GRCh38
1394780 c.81G>A p.S27S Chr.19: 46494713-46494713 on Build GRCh38
1612443 c.118G>A p.A40T Chr.19: 46494750-46494750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967991080 Chr.19: 46494801-46494801 on Build GRCh38
rs771224713 Chr.19: 46494717-46494717 on Build GRCh38
rs919978202 Chr.19: 46494800-46494800 on Build GRCh38
rs754635701 Chr.19: 46494678-46494678 on Build GRCh38
rs767184857 Chr.19: 46494826-46494826 on Build GRCh38
rs772337171 Chr.19: 46494748-46494748 on Build GRCh38
rs780606470 Chr.19: 46494686-46494686 on Build GRCh38
rs146376571 Chr.19: 46494774-46494774 on Build GRCh38
rs760855001 Chr.19: 46494759-46494759 on Build GRCh38
rs375528723 Chr.19: 46494728-46494728 on Build GRCh38
rs762932360 Chr.19: 46494778-46494778 on Build GRCh38
rs751190548 Chr.19: 46494677-46494677 on Build GRCh38
rs758269236 Chr.19: 46494642-46494642 on Build GRCh38
rs775495148 Chr.19: 46494756-46494756 on Build GRCh38
rs138594215 Chr.19: 46494715-46494715 on Build GRCh38
rs1025320269 Chr.19: 46494707-46494707 on Build GRCh38
rs751337008 Chr.19: 46494812-46494812 on Build GRCh38
rs764199146 Chr.19: 46494766-46494766 on Build GRCh38
rs142336941 Chr.19: 46494721-46494721 on Build GRCh38
rs1032034744 Chr.19: 46494670-46494670 on Build GRCh38
rs143805999 Chr.19: 46494647-46494647 on Build GRCh38
rs766133213 Chr.19: 46494789-46494789 on Build GRCh38
rs139093853 Chr.19: 46494836-46494836 on Build GRCh38
rs35793066 Chr.19: 46494677-46494678 on Build GRCh38
rs372340134 Chr.19: 46494714-46494714 on Build GRCh38
rs752288047 Chr.19: 46494829-46494829 on Build GRCh38
rs754651152 Chr.19: 46494820-46494820 on Build GRCh38
rs747653196 Chr.19: 46494688-46494688 on Build GRCh38
rs1022186470 Chr.19: 46494781-46494781 on Build GRCh38
rs1012007352 Chr.19: 46494702-46494702 on Build GRCh38

View Full Product Details