Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151700628-151700889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRG3
Entrez Gene ID
79057
Gene Name
proline rich and Gla domain 3
Gene Aliases
PRGP3, TMG3
UniGene
Hs.209253
Gene Chromosome Location
Chr.X: 151694606-151701591 on Build GRCh38
Transcripts
NM_024082.3 NR_033262.1 XM_005274747.3 XM_011531197.2

SNP Detail

SNP ID Chromosome Location
rs769628736 Chr.X: 151700843-151700843 on Build GRCh38
rs750961776 Chr.X: 151700780-151700780 on Build GRCh38
rs750828642 Chr.X: 151700711-151700711 on Build GRCh38
rs764746380 Chr.X: 151700824-151700824 on Build GRCh38
rs770746370 Chr.X: 151700750-151700750 on Build GRCh38
rs370201889 Chr.X: 151700859-151700859 on Build GRCh38
rs756248851 Chr.X: 151700678-151700678 on Build GRCh38
rs1026297893 Chr.X: 151700713-151700713 on Build GRCh38
rs770997065 Chr.X: 151700831-151700831 on Build GRCh38
rs866763040 Chr.X: 151700731-151700731 on Build GRCh38
rs765014529 Chr.X: 151700666-151700666 on Build GRCh38
rs760934250 Chr.X: 151700848-151700848 on Build GRCh38
rs965372200 Chr.X: 151700757-151700757 on Build GRCh38
rs746839699 Chr.X: 151700743-151700743 on Build GRCh38
rs771392906 Chr.X: 151700850-151700850 on Build GRCh38
rs1017202575 Chr.X: 151700698-151700698 on Build GRCh38
rs776874333 Chr.X: 151700851-151700851 on Build GRCh38
rs142444421 Chr.X: 151700801-151700801 on Build GRCh38
rs989647414 Chr.X: 151700770-151700770 on Build GRCh38
rs758007267 Chr.X: 151700826-151700826 on Build GRCh38
rs138720379 Chr.X: 151700660-151700660 on Build GRCh38
rs772083345 Chr.X: 151700820-151700820 on Build GRCh38
rs1017849399 Chr.X: 151700730-151700730 on Build GRCh38
rs778416070 Chr.X: 151700727-151700727 on Build GRCh38
rs775395243 Chr.X: 151700697-151700697 on Build GRCh38
rs778667847 Chr.X: 151700669-151700669 on Build GRCh38
rs895088635 Chr.X: 151700685-151700685 on Build GRCh38
rs767922631 Chr.X: 151700710-151700710 on Build GRCh38
rs139807152 Chr.X: 151700825-151700825 on Build GRCh38
rs200265801 Chr.X: 151700782-151700782 on Build GRCh38
rs143153628 Chr.X: 151700707-151700707 on Build GRCh38
rs779085260 Chr.X: 151700844-151700844 on Build GRCh38
rs776529242 Chr.X: 151700866-151700866 on Build GRCh38
rs780990958 Chr.X: 151700840-151700840 on Build GRCh38
rs150201138 Chr.X: 151700655-151700655 on Build GRCh38
rs746893048 Chr.X: 151700682-151700682 on Build GRCh38
rs939232157 Chr.X: 151700676-151700676 on Build GRCh38
rs752173615 Chr.X: 151700865-151700865 on Build GRCh38
rs143276868 Chr.X: 151700845-151700845 on Build GRCh38
rs199718169 Chr.X: 151700792-151700792 on Build GRCh38
rs373242051 Chr.X: 151700729-151700729 on Build GRCh38
rs747872268 Chr.X: 151700674-151700674 on Build GRCh38
rs765695965 Chr.X: 151700864-151700864 on Build GRCh38
rs763136904 Chr.X: 151700702-151700702 on Build GRCh38
rs748526759 Chr.X: 151700824-151700825 on Build GRCh38
rs377405021 Chr.X: 151700853-151700853 on Build GRCh38
rs143962455 Chr.X: 151700758-151700758 on Build GRCh38
rs780375684 Chr.X: 151700783-151700783 on Build GRCh38
rs779322904 Chr.X: 151700666-151700672 on Build GRCh38
rs375409446 Chr.X: 151700841-151700841 on Build GRCh38
rs150521353 Chr.X: 151700808-151700808 on Build GRCh38
rs377331866 Chr.X: 151700786-151700786 on Build GRCh38
rs1008897909 Chr.X: 151700860-151700860 on Build GRCh38
rs772442871 Chr.X: 151700793-151700793 on Build GRCh38
rs761342209 Chr.X: 151700722-151700722 on Build GRCh38
rs751336477 Chr.X: 151700829-151700829 on Build GRCh38
rs147473030 Chr.X: 151700712-151700712 on Build GRCh38
rs4323608 Chr.X: 151700795-151700795 on Build GRCh38
rs755150330 Chr.X: 151700799-151700799 on Build GRCh38
rs751033642 Chr.X: 151700652-151700652 on Build GRCh38
rs369580690 Chr.X: 151700692-151700692 on Build GRCh38
rs12394468 Chr.X: 151700846-151700846 on Build GRCh38
rs1019184963 Chr.X: 151700708-151700708 on Build GRCh38
rs772996429 Chr.X: 151700679-151700679 on Build GRCh38
rs762259905 Chr.X: 151700709-151700709 on Build GRCh38
rs766939423 Chr.X: 151700714-151700714 on Build GRCh38
rs138478062 Chr.X: 151700719-151700719 on Build GRCh38
rs950659306 Chr.X: 151700693-151700693 on Build GRCh38
rs768041636 Chr.X: 151700728-151700728 on Build GRCh38
rs746864658 Chr.X: 151700828-151700828 on Build GRCh38
rs895042335 Chr.X: 151700818-151700818 on Build GRCh38

View Full Product Details