Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64481655-64481924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBK1
Entrez Gene ID
29110
Gene Name
TANK binding kinase 1
Gene Aliases
FTDALS4, NAK, T2K
UniGene
Hs.505874
Gene Chromosome Location
Chr.12: 64452060-64502119 on Build GRCh38
Transcripts
NM_013254.3 XM_005268809.1 XM_005268810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797744 c.822G>T p.Q274H Chr.12: 64481851-64481851 on Build GRCh38
5841714 c.862G>A p.D288N Chr.12: 64481891-64481891 on Build GRCh38
1228645 c.866A>G p.Q289R Chr.12: 64481895-64481895 on Build GRCh38
942514 c.841C>A p.L281I Chr.12: 64481870-64481870 on Build GRCh38
381896 c.813G>T p.R271R Chr.12: 64481842-64481842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376075252 Chr.12: 64481835-64481835 on Build GRCh38
rs931538116 Chr.12: 64481749-64481749 on Build GRCh38
rs767857968 Chr.12: 64481805-64481805 on Build GRCh38
rs757938202 Chr.12: 64481845-64481845 on Build GRCh38
rs11837054 Chr.12: 64481769-64481769 on Build GRCh38
rs878939532 Chr.12: 64481834-64481834 on Build GRCh38
rs918795970 Chr.12: 64481735-64481735 on Build GRCh38
rs369821427 Chr.12: 64481809-64481809 on Build GRCh38
rs34774243 Chr.12: 64481900-64481900 on Build GRCh38
rs534626168 Chr.12: 64481787-64481787 on Build GRCh38
rs557248505 Chr.12: 64481772-64481772 on Build GRCh38
rs781034590 Chr.12: 64481898-64481898 on Build GRCh38
rs764851412 Chr.12: 64481817-64481817 on Build GRCh38
rs371757662 Chr.12: 64481802-64481802 on Build GRCh38
rs763504116 Chr.12: 64481688-64481688 on Build GRCh38
rs568091990 Chr.12: 64481684-64481684 on Build GRCh38
rs935495819 Chr.12: 64481753-64481753 on Build GRCh38
rs754873844 Chr.12: 64481884-64481884 on Build GRCh38
rs751481440 Chr.12: 64481857-64481857 on Build GRCh38
rs905184241 Chr.12: 64481858-64481858 on Build GRCh38
rs144982345 Chr.12: 64481773-64481773 on Build GRCh38
rs979646277 Chr.12: 64481723-64481723 on Build GRCh38
rs1023995234 Chr.12: 64481710-64481710 on Build GRCh38
rs753073799 Chr.12: 64481806-64481806 on Build GRCh38
rs961676287 Chr.12: 64481721-64481721 on Build GRCh38
rs551774139 Chr.12: 64481820-64481820 on Build GRCh38
rs946010358 Chr.12: 64481818-64481818 on Build GRCh38
rs540025705 Chr.12: 64481775-64481775 on Build GRCh38
rs925578225 Chr.12: 64481750-64481750 on Build GRCh38
rs774792491 Chr.12: 64481791-64481791 on Build GRCh38

View Full Product Details