Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237566983-237567208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRLH
Entrez Gene ID
51052
Gene Name
prolactin releasing hormone
Gene Aliases
PRH, PRRP
UniGene
Hs.247710
Gene Chromosome Location
Chr.2: 237566574-237567175 on Build GRCh38
Transcripts
NM_015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669888 c.164G>A p.R55Q Chr.2: 237567075-237567075 on Build GRCh38
1724621 c.106G>A p.D36N Chr.2: 237567017-237567017 on Build GRCh38
1018386 c.136G>A p.G46R Chr.2: 237567047-237567047 on Build GRCh38
6157478 c.111C>T p.I37I Chr.2: 237567022-237567022 on Build GRCh38
1018387 c.206G>A p.R69Q Chr.2: 237567117-237567117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771093364 Chr.2: 237567028-237567028 on Build GRCh38
rs766419451 Chr.2: 237567060-237567060 on Build GRCh38
rs141578550 Chr.2: 237567046-237567046 on Build GRCh38
rs774491644 Chr.2: 237567134-237567134 on Build GRCh38
rs989594408 Chr.2: 237567037-237567043 on Build GRCh38
rs781113806 Chr.2: 237567015-237567016 on Build GRCh38
rs553942767 Chr.2: 237567143-237567143 on Build GRCh38
rs748695228 Chr.2: 237567063-237567063 on Build GRCh38
rs779919303 Chr.2: 237567121-237567121 on Build GRCh38
rs750107491 Chr.2: 237567090-237567090 on Build GRCh38
rs771617955 Chr.2: 237567137-237567137 on Build GRCh38
rs753007054 Chr.2: 237567044-237567045 on Build GRCh38
rs746911034 Chr.2: 237567090-237567090 on Build GRCh38
rs746063425 Chr.2: 237567072-237567072 on Build GRCh38
rs758740474 Chr.2: 237567098-237567098 on Build GRCh38
rs760398931 Chr.2: 237567147-237567147 on Build GRCh38
rs753457968 Chr.2: 237567150-237567150 on Build GRCh38
rs139168075 Chr.2: 237567116-237567116 on Build GRCh38
rs748592104 Chr.2: 237567016-237567016 on Build GRCh38
rs372593695 Chr.2: 237567075-237567075 on Build GRCh38
rs750043248 Chr.2: 237567058-237567058 on Build GRCh38
rs112601669 Chr.2: 237567183-237567183 on Build GRCh38
rs775553885 Chr.2: 237567040-237567040 on Build GRCh38
rs757385411 Chr.2: 237567111-237567111 on Build GRCh38
rs148681723 Chr.2: 237567099-237567099 on Build GRCh38
rs935419737 Chr.2: 237567084-237567084 on Build GRCh38
rs144703716 Chr.2: 237567117-237567117 on Build GRCh38
rs138895198 Chr.2: 237567065-237567065 on Build GRCh38
rs755432010 Chr.2: 237567059-237567059 on Build GRCh38
rs938323018 Chr.2: 237567049-237567049 on Build GRCh38
rs773211005 Chr.2: 237567026-237567026 on Build GRCh38
rs774405650 Chr.2: 237567081-237567081 on Build GRCh38
rs764429291 Chr.2: 237567158-237567158 on Build GRCh38
rs112622920 Chr.2: 237567038-237567038 on Build GRCh38
rs761789426 Chr.2: 237567135-237567135 on Build GRCh38
rs373931155 Chr.2: 237567037-237567037 on Build GRCh38
rs777852593 Chr.2: 237567064-237567064 on Build GRCh38
rs749139542 Chr.2: 237567123-237567123 on Build GRCh38
rs146226371 Chr.2: 237567093-237567093 on Build GRCh38
rs1035275151 Chr.2: 237567149-237567149 on Build GRCh38
rs377353869 Chr.2: 237567110-237567110 on Build GRCh38
rs763373473 Chr.2: 237567155-237567155 on Build GRCh38
rs781544659 Chr.2: 237567071-237567071 on Build GRCh38
rs568386891 Chr.2: 237567079-237567079 on Build GRCh38
rs760779070 Chr.2: 237567074-237567074 on Build GRCh38
rs768537215 Chr.2: 237567129-237567129 on Build GRCh38
rs370035806 Chr.2: 237567066-237567066 on Build GRCh38
rs773361810 Chr.2: 237567044-237567044 on Build GRCh38
rs757636413 Chr.2: 237567184-237567184 on Build GRCh38
rs761485193 Chr.2: 237567082-237567082 on Build GRCh38
rs776696083 Chr.2: 237567035-237567035 on Build GRCh38
rs766927745 Chr.2: 237567087-237567087 on Build GRCh38
rs142015444 Chr.2: 237567062-237567062 on Build GRCh38
rs999502941 Chr.2: 237567112-237567112 on Build GRCh38
rs772156240 Chr.2: 237567022-237567022 on Build GRCh38
rs753013498 Chr.2: 237567097-237567097 on Build GRCh38
rs202225879 Chr.2: 237567027-237567027 on Build GRCh38
rs376015494 Chr.2: 237567045-237567045 on Build GRCh38
rs147108124 Chr.2: 237567047-237567047 on Build GRCh38
rs886981117 Chr.2: 237567151-237567151 on Build GRCh38
rs200614877 Chr.2: 237567148-237567148 on Build GRCh38
rs145946342 Chr.2: 237567094-237567094 on Build GRCh38
rs769865345 Chr.2: 237567120-237567120 on Build GRCh38
rs370516153 Chr.2: 237567095-237567095 on Build GRCh38
rs372698942 Chr.2: 237567107-237567107 on Build GRCh38

View Full Product Details