Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33147149-33147388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN2
Entrez Gene ID
113451
Gene Name
antizyme inhibitor 2
Gene Aliases
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp
UniGene
Hs.101807
Gene Chromosome Location
Chr.1: 33081110-33162288 on Build GRCh38
Transcripts
XM_017000167.1 XM_017000174.1 NM_018207.2 XM_011541705.2 XM_017001629.1 XM_017001630.1

Gene Symbol
TRIM62
Entrez Gene ID
55223
Gene Name
tripartite motif containing 62
Gene Aliases
DEAR1
UniGene
Hs.656006
Gene Chromosome Location
Chr.1: 33144603-33184774 on Build GRCh38
Transcripts
XM_017000167.1 XM_017000174.1 NM_018207.2 XM_011541705.2 XM_017001629.1 XM_017001630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4736198 c.1217G>A p.R406Q Chr.1: 33147388-33147388 on Build GRCh38
5565963 c.1228C>T p.R410W Chr.1: 33147377-33147377 on Build GRCh38
1683127 c.1247delT p.V416fs*>60 Chr.1: 33147358-33147358 on Build GRCh38
3782464 c.1245G>T p.K415N Chr.1: 33147360-33147360 on Build GRCh38
4031578 c.1422C>T p.R474R Chr.1: 33147183-33147183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911108993 Chr.1: 33147247-33147250 on Build GRCh38
rs369476406 Chr.1: 33147273-33147273 on Build GRCh38
rs759270033 Chr.1: 33147264-33147264 on Build GRCh38
rs142450553 Chr.1: 33147210-33147210 on Build GRCh38
rs41310426 Chr.1: 33147226-33147226 on Build GRCh38
rs746662481 Chr.1: 33147279-33147279 on Build GRCh38
rs774201238 Chr.1: 33147228-33147228 on Build GRCh38
rs780069356 Chr.1: 33147291-33147291 on Build GRCh38
rs747388545 Chr.1: 33147301-33147301 on Build GRCh38
rs755449501 Chr.1: 33147317-33147317 on Build GRCh38
rs966425324 Chr.1: 33147189-33147189 on Build GRCh38
rs776011218 Chr.1: 33147358-33147358 on Build GRCh38
rs142555750 Chr.1: 33147199-33147199 on Build GRCh38
rs772577078 Chr.1: 33147204-33147204 on Build GRCh38
rs772814762 Chr.1: 33147337-33147337 on Build GRCh38
rs372526245 Chr.1: 33147278-33147278 on Build GRCh38
rs780488792 Chr.1: 33147184-33147184 on Build GRCh38
rs748649754 Chr.1: 33147198-33147198 on Build GRCh38
rs754617018 Chr.1: 33147330-33147330 on Build GRCh38
rs781668478 Chr.1: 33147324-33147324 on Build GRCh38
rs750420902 Chr.1: 33147280-33147280 on Build GRCh38
rs770155503 Chr.1: 33147336-33147336 on Build GRCh38
rs375728015 Chr.1: 33147302-33147302 on Build GRCh38
rs758674028 Chr.1: 33147281-33147281 on Build GRCh38
rs202210162 Chr.1: 33147339-33147339 on Build GRCh38
rs61758110 Chr.1: 33147227-33147227 on Build GRCh38
rs61737100 Chr.1: 33147188-33147188 on Build GRCh38
rs143361624 Chr.1: 33147272-33147272 on Build GRCh38
rs769287444 Chr.1: 33147185-33147185 on Build GRCh38
rs774049265 Chr.1: 33147355-33147355 on Build GRCh38
rs770630617 Chr.1: 33147344-33147344 on Build GRCh38

View Full Product Details