Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 96334120-96334359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F2
Entrez Gene ID
7026
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
UniGene
Hs.347991
Gene Chromosome Location
Chr.15: 96325928-96340263 on Build GRCh38
Transcripts
NM_001145155.1 NM_001145156.1 NM_001145157.1 NM_021005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4058059 c.260T>C p.L87P Chr.15: 96334292-96334292 on Build GRCh38
5734542 c.647C>T p.A216V Chr.15: 96334280-96334280 on Build GRCh38
6078450 c.274G>T p.E92* Chr.15: 96334306-96334306 on Build GRCh38
4058058 c.659T>C p.L220P Chr.15: 96334292-96334292 on Build GRCh38
4255090 c.96G>A p.A32A Chr.15: 96334128-96334128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991918103 Chr.15: 96334176-96334176 on Build GRCh38
rs587777372 Chr.15: 96334247-96334247 on Build GRCh38
rs143951395 Chr.15: 96334170-96334170 on Build GRCh38
rs745505294 Chr.15: 96334266-96334266 on Build GRCh38
rs374919946 Chr.15: 96334317-96334317 on Build GRCh38
rs756760829 Chr.15: 96334239-96334239 on Build GRCh38
rs780916977 Chr.15: 96334249-96334249 on Build GRCh38
rs762857147 Chr.15: 96334143-96334143 on Build GRCh38
rs758719602 Chr.15: 96334227-96334227 on Build GRCh38
rs141514117 Chr.15: 96334329-96334329 on Build GRCh38
rs765458532 Chr.15: 96334191-96334191 on Build GRCh38
rs545799827 Chr.15: 96334155-96334155 on Build GRCh38
rs749054602 Chr.15: 96334283-96334283 on Build GRCh38
rs754051151 Chr.15: 96334175-96334175 on Build GRCh38
rs769518278 Chr.15: 96334267-96334267 on Build GRCh38
rs778295872 Chr.15: 96334230-96334230 on Build GRCh38
rs747521046 Chr.15: 96334235-96334235 on Build GRCh38
rs761719602 Chr.15: 96334302-96334302 on Build GRCh38
rs148228212 Chr.15: 96334320-96334320 on Build GRCh38
rs143464692 Chr.15: 96334147-96334147 on Build GRCh38
rs927149153 Chr.15: 96334213-96334213 on Build GRCh38
rs768381736 Chr.15: 96334284-96334284 on Build GRCh38
rs772477460 Chr.15: 96334242-96334244 on Build GRCh38
rs544461724 Chr.15: 96334281-96334281 on Build GRCh38
rs1036089807 Chr.15: 96334151-96334151 on Build GRCh38
rs531461667 Chr.15: 96334212-96334212 on Build GRCh38
rs765401593 Chr.15: 96334335-96334335 on Build GRCh38
rs774311305 Chr.15: 96334290-96334290 on Build GRCh38
rs370572235 Chr.15: 96334161-96334161 on Build GRCh38
rs968967821 Chr.15: 96334260-96334260 on Build GRCh38
rs755250396 Chr.15: 96334189-96334189 on Build GRCh38

View Full Product Details