Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55545484-55545729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBK3
Entrez Gene ID
100130827
Gene Name
SH3 domain binding kinase family member 3
Gene Aliases
SGK110
UniGene
Hs.712129
Gene Chromosome Location
Chr.19: 55540558-55545543 on Build GRCh38
Transcripts
NM_001199824.2

SNP Detail

SNP ID Chromosome Location
rs536250447 Chr.19: 55545610-55545610 on Build GRCh38
rs369829845 Chr.19: 55545703-55545703 on Build GRCh38
rs866806400 Chr.19: 55545647-55545647 on Build GRCh38
rs1054209279 Chr.19: 55545648-55545648 on Build GRCh38
rs954345867 Chr.19: 55545620-55545620 on Build GRCh38
rs998768210 Chr.19: 55545596-55545600 on Build GRCh38
rs938689619 Chr.19: 55545702-55545702 on Build GRCh38
rs375142265 Chr.19: 55545578-55545578 on Build GRCh38
rs893836207 Chr.19: 55545656-55545656 on Build GRCh38
rs35384972 Chr.19: 55545581-55545582 on Build GRCh38
rs188492399 Chr.19: 55545588-55545588 on Build GRCh38
rs571837455 Chr.19: 55545531-55545531 on Build GRCh38
rs573017495 Chr.19: 55545663-55545663 on Build GRCh38
rs376560321 Chr.19: 55545638-55545638 on Build GRCh38
rs1047089211 Chr.19: 55545525-55545525 on Build GRCh38
rs34882755 Chr.19: 55545503-55545503 on Build GRCh38
rs975618691 Chr.19: 55545658-55545658 on Build GRCh38
rs191810173 Chr.19: 55545589-55545589 on Build GRCh38
rs538783191 Chr.19: 55545665-55545665 on Build GRCh38
rs1038892073 Chr.19: 55545616-55545616 on Build GRCh38
rs371980262 Chr.19: 55545605-55545605 on Build GRCh38
rs931468881 Chr.19: 55545681-55545681 on Build GRCh38
rs776310046 Chr.19: 55545521-55545521 on Build GRCh38
rs866302035 Chr.19: 55545580-55545580 on Build GRCh38
rs996778092 Chr.19: 55545643-55545643 on Build GRCh38
rs1047496368 Chr.19: 55545583-55545583 on Build GRCh38
rs553022886 Chr.19: 55545642-55545642 on Build GRCh38
rs757953959 Chr.19: 55545573-55545574 on Build GRCh38
rs184332753 Chr.19: 55545689-55545689 on Build GRCh38
rs781726824 Chr.19: 55545577-55545577 on Build GRCh38
rs973069593 Chr.19: 55545520-55545520 on Build GRCh38
rs150849351 Chr.19: 55545688-55545688 on Build GRCh38
rs894569558 Chr.19: 55545582-55545582 on Build GRCh38
rs764769521 Chr.19: 55545565-55545565 on Build GRCh38
rs567515457 Chr.19: 55545593-55545593 on Build GRCh38
rs765919952 Chr.19: 55545516-55545516 on Build GRCh38

View Full Product Details