Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35625304-35625529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGAPA1
Entrez Gene ID
253959
Gene Name
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
UniGene
Hs.113150
Gene Chromosome Location
Chr.14: 35538352-35809304 on Build GRCh38
Transcripts
NM_001283043.1 NM_001283044.1 NM_014990.1 NM_194301.2 XM_005267491.3 XM_005267492.3 XM_005267493.3 XM_005267498.4 XM_006720098.2 XM_006720099.2 XM_006720100.3 XM_006720101.2 XM_006720102.3 XM_006720103.2 XM_011536615.2 XM_017021141.1 XM_017021142.1 XM_017021143.1 XM_017021144.1 XM_017021145.1 XM_017021146.1 XM_017021147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986295 c.5405G>C p.R1802T Chr.14: 35625367-35625367 on Build GRCh38
1685708 c.5396_5397insT p.L1799fs*19 Chr.14: 35625375-35625376 on Build GRCh38
469931 c.5384A>G p.E1795G Chr.14: 35625388-35625388 on Build GRCh38
1685707 c.5396_5397insT p.L1799fs*19 Chr.14: 35625375-35625376 on Build GRCh38
348929 c.5358G>A p.L1786L Chr.14: 35625414-35625414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755520555 Chr.14: 35625434-35625437 on Build GRCh38
rs761204814 Chr.14: 35625466-35625466 on Build GRCh38
rs528899646 Chr.14: 35625443-35625443 on Build GRCh38
rs756247627 Chr.14: 35625404-35625404 on Build GRCh38
rs977675739 Chr.14: 35625485-35625485 on Build GRCh38
rs189026062 Chr.14: 35625438-35625438 on Build GRCh38
rs767906488 Chr.14: 35625337-35625337 on Build GRCh38
rs752942621 Chr.14: 35625477-35625477 on Build GRCh38
rs967330479 Chr.14: 35625482-35625483 on Build GRCh38
rs538008773 Chr.14: 35625493-35625493 on Build GRCh38
rs200497444 Chr.14: 35625454-35625454 on Build GRCh38
rs373859527 Chr.14: 35625338-35625338 on Build GRCh38
rs780083892 Chr.14: 35625398-35625398 on Build GRCh38
rs192294732 Chr.14: 35625492-35625492 on Build GRCh38
rs140958336 Chr.14: 35625382-35625382 on Build GRCh38
rs376831497 Chr.14: 35625472-35625472 on Build GRCh38
rs879593746 Chr.14: 35625366-35625366 on Build GRCh38
rs1047564031 Chr.14: 35625442-35625442 on Build GRCh38
rs772318351 Chr.14: 35625450-35625450 on Build GRCh38
rs906409128 Chr.14: 35625377-35625377 on Build GRCh38
rs548775734 Chr.14: 35625480-35625480 on Build GRCh38
rs200815371 Chr.14: 35625414-35625414 on Build GRCh38
rs749517987 Chr.14: 35625427-35625427 on Build GRCh38
rs369238925 Chr.14: 35625396-35625396 on Build GRCh38
rs751451957 Chr.14: 35625399-35625399 on Build GRCh38

View Full Product Details