Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73088284-73088511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP5
Entrez Gene ID
26056
Gene Name
RAB11 family interacting protein 5
Gene Aliases
GAF1, RIP11, pp75
UniGene
Hs.24557
Gene Chromosome Location
Chr.2: 73073382-73113018 on Build GRCh38
Transcripts
NM_015470.2 XM_005264251.3 XM_005264252.3 XM_005264253.4 XM_006711985.3 XM_011532753.2 XM_011532754.2 XM_011532755.2 XM_011532756.2 XM_017003788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841259 c.1142G>A p.R381H Chr.2: 73088476-73088476 on Build GRCh38
3583106 c.1141C>T p.R381C Chr.2: 73088477-73088477 on Build GRCh38
6893226 c.1257C>T p.H419H Chr.2: 73088361-73088361 on Build GRCh38
460996 c.1218G>C p.L406L Chr.2: 73088400-73088400 on Build GRCh38
4624858 c.1115C>T p.S372F Chr.2: 73088503-73088503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150166054 Chr.2: 73088473-73088473 on Build GRCh38
rs747892372 Chr.2: 73088401-73088401 on Build GRCh38
rs202050000 Chr.2: 73088489-73088489 on Build GRCh38
rs138135562 Chr.2: 73088407-73088407 on Build GRCh38
rs760273170 Chr.2: 73088404-73088404 on Build GRCh38
rs759086154 Chr.2: 73088490-73088490 on Build GRCh38
rs958242250 Chr.2: 73088310-73088310 on Build GRCh38
rs760021988 Chr.2: 73088421-73088421 on Build GRCh38
rs764749815 Chr.2: 73088491-73088491 on Build GRCh38
rs746014225 Chr.2: 73088466-73088466 on Build GRCh38
rs145839919 Chr.2: 73088333-73088333 on Build GRCh38
rs971289714 Chr.2: 73088347-73088347 on Build GRCh38
rs760277863 Chr.2: 73088318-73088318 on Build GRCh38
rs963183027 Chr.2: 73088367-73088367 on Build GRCh38
rs765979365 Chr.2: 73088473-73088484 on Build GRCh38
rs768686792 Chr.2: 73088468-73088468 on Build GRCh38
rs768774709 Chr.2: 73088346-73088346 on Build GRCh38
rs561703908 Chr.2: 73088338-73088338 on Build GRCh38
rs753028183 Chr.2: 73088436-73088436 on Build GRCh38
rs758741352 Chr.2: 73088443-73088443 on Build GRCh38
rs773792064 Chr.2: 73088489-73088495 on Build GRCh38
rs573589936 Chr.2: 73088438-73088438 on Build GRCh38
rs1023911671 Chr.2: 73088341-73088341 on Build GRCh38
rs536535862 Chr.2: 73088339-73088339 on Build GRCh38
rs767270562 Chr.2: 73088303-73088303 on Build GRCh38
rs1001677387 Chr.2: 73088360-73088360 on Build GRCh38
rs765846322 Chr.2: 73088409-73088409 on Build GRCh38
rs779188928 Chr.2: 73088323-73088323 on Build GRCh38
rs757435446 Chr.2: 73088449-73088449 on Build GRCh38
rs749873719 Chr.2: 73088306-73088306 on Build GRCh38
rs772885269 Chr.2: 73088486-73088486 on Build GRCh38
rs369090487 Chr.2: 73088350-73088350 on Build GRCh38
rs199517071 Chr.2: 73088476-73088476 on Build GRCh38
rs569691441 Chr.2: 73088444-73088444 on Build GRCh38
rs140983118 Chr.2: 73088328-73088328 on Build GRCh38
rs753357623 Chr.2: 73088411-73088411 on Build GRCh38
rs765798618 Chr.2: 73088430-73088430 on Build GRCh38
rs747218615 Chr.2: 73088336-73088336 on Build GRCh38
rs771856878 Chr.2: 73088477-73088477 on Build GRCh38
rs781409741 Chr.2: 73088453-73088453 on Build GRCh38
rs994434150 Chr.2: 73088403-73088403 on Build GRCh38
rs772868278 Chr.2: 73088382-73088382 on Build GRCh38
rs761712902 Chr.2: 73088365-73088365 on Build GRCh38
rs754300332 Chr.2: 73088321-73088321 on Build GRCh38
rs762617959 Chr.2: 73088348-73088348 on Build GRCh38
rs144951768 Chr.2: 73088342-73088342 on Build GRCh38
rs753117895 Chr.2: 73088325-73088325 on Build GRCh38
rs771598578 Chr.2: 73088370-73088370 on Build GRCh38

View Full Product Details