Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63698325-63698570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTEL1-TNFRSF6B
Entrez Gene ID
100533107
Gene Name
RTEL1-TNFRSF6B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.745057
Gene Chromosome Location
Chr.20: 63657810-63698698 on Build GRCh38
Transcripts
NR_037882.1 NM_003823.3

Gene Symbol
TNFRSF6B
Entrez Gene ID
8771
Gene Name
TNF receptor superfamily member 6b
Gene Aliases
DCR3, DJ583P15.1.1, M68, M68E, TR6
UniGene
Hs.434878
Gene Chromosome Location
Chr.20: 63696651-63698698 on Build GRCh38
Transcripts
NR_037882.1 NM_003823.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734901 c.812C>T p.A271V Chr.20: 63698472-63698472 on Build GRCh38
6424034 c.850A>T p.R284W Chr.20: 63698510-63698510 on Build GRCh38
4734902 c.874G>A p.V292I Chr.20: 63698534-63698534 on Build GRCh38
6424422 c.808G>T p.G270W Chr.20: 63698468-63698468 on Build GRCh38
4734903 c.893C>T p.P298L Chr.20: 63698553-63698553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377030676 Chr.20: 63698409-63698409 on Build GRCh38
rs1041805018 Chr.20: 63698460-63698460 on Build GRCh38
rs147543198 Chr.20: 63698352-63698352 on Build GRCh38
rs546975323 Chr.20: 63698472-63698472 on Build GRCh38
rs755869527 Chr.20: 63698402-63698402 on Build GRCh38
rs768990640 Chr.20: 63698382-63698382 on Build GRCh38
rs771489318 Chr.20: 63698461-63698461 on Build GRCh38
rs772995877 Chr.20: 63698510-63698510 on Build GRCh38
rs370956731 Chr.20: 63698416-63698416 on Build GRCh38
rs750669514 Chr.20: 63698385-63698385 on Build GRCh38
rs767090761 Chr.20: 63698433-63698433 on Build GRCh38
rs143300172 Chr.20: 63698351-63698351 on Build GRCh38
rs757539968 Chr.20: 63698488-63698488 on Build GRCh38
rs754828981 Chr.20: 63698360-63698360 on Build GRCh38
rs772115276 Chr.20: 63698373-63698373 on Build GRCh38
rs755157676 Chr.20: 63698411-63698411 on Build GRCh38
rs781286204 Chr.20: 63698447-63698447 on Build GRCh38
rs769716653 Chr.20: 63698548-63698548 on Build GRCh38
rs937966789 Chr.20: 63698412-63698412 on Build GRCh38
rs530751226 Chr.20: 63698393-63698393 on Build GRCh38
rs770649191 Chr.20: 63698420-63698420 on Build GRCh38
rs748271642 Chr.20: 63698449-63698449 on Build GRCh38
rs923508643 Chr.20: 63698381-63698381 on Build GRCh38
rs752764747 Chr.20: 63698481-63698481 on Build GRCh38
rs751848480 Chr.20: 63698446-63698446 on Build GRCh38
rs375903710 Chr.20: 63698379-63698379 on Build GRCh38
rs779650602 Chr.20: 63698537-63698537 on Build GRCh38
rs767910637 Chr.20: 63698384-63698395 on Build GRCh38
rs756317669 Chr.20: 63698483-63698483 on Build GRCh38
rs749673648 Chr.20: 63698419-63698419 on Build GRCh38
rs950972975 Chr.20: 63698463-63698463 on Build GRCh38
rs769653561 Chr.20: 63698504-63698504 on Build GRCh38
rs548373058 Chr.20: 63698368-63698368 on Build GRCh38
rs781192378 Chr.20: 63698361-63698361 on Build GRCh38
rs779100044 Chr.20: 63698369-63698369 on Build GRCh38
rs566823245 Chr.20: 63698484-63698484 on Build GRCh38
rs766159208 Chr.20: 63698392-63698392 on Build GRCh38
rs753574890 Chr.20: 63698438-63698438 on Build GRCh38
rs909306336 Chr.20: 63698400-63698400 on Build GRCh38
rs762667464 Chr.20: 63698511-63698511 on Build GRCh38
rs750168237 Chr.20: 63698346-63698346 on Build GRCh38
rs748740001 Chr.20: 63698414-63698414 on Build GRCh38
rs757741103 Chr.20: 63698529-63698529 on Build GRCh38
rs555469304 Chr.20: 63698533-63698533 on Build GRCh38
rs541025487 Chr.20: 63698497-63698497 on Build GRCh38
rs746126394 Chr.20: 63698371-63698371 on Build GRCh38
rs917853575 Chr.20: 63698519-63698519 on Build GRCh38
rs780272290 Chr.20: 63698410-63698410 on Build GRCh38
rs767763294 Chr.20: 63698515-63698515 on Build GRCh38
rs759416218 Chr.20: 63698531-63698532 on Build GRCh38
rs745580105 Chr.20: 63698466-63698466 on Build GRCh38
rs765128731 Chr.20: 63698473-63698473 on Build GRCh38
rs570762078 Chr.20: 63698432-63698432 on Build GRCh38
rs369340699 Chr.20: 63698435-63698435 on Build GRCh38
rs778395396 Chr.20: 63698462-63698462 on Build GRCh38
rs1017774705 Chr.20: 63698363-63698363 on Build GRCh38
rs561847351 Chr.20: 63698391-63698391 on Build GRCh38
rs750208461 Chr.20: 63698474-63698474 on Build GRCh38
rs753653499 Chr.20: 63698408-63698408 on Build GRCh38
rs369558371 Chr.20: 63698390-63698390 on Build GRCh38
rs761928512 Chr.20: 63698356-63698380 on Build GRCh38
rs752376808 Chr.20: 63698364-63698364 on Build GRCh38
rs779286455 Chr.20: 63698491-63698491 on Build GRCh38
rs372049533 Chr.20: 63698468-63698468 on Build GRCh38
rs759098640 Chr.20: 63698395-63698395 on Build GRCh38
rs759092822 Chr.20: 63698527-63698527 on Build GRCh38
rs749655628 Chr.20: 63698456-63698456 on Build GRCh38
rs758758173 Chr.20: 63698534-63698534 on Build GRCh38
rs780071498 Chr.20: 63698376-63698376 on Build GRCh38
rs760801972 Chr.20: 63698518-63698518 on Build GRCh38
rs766509761 Chr.20: 63698445-63698445 on Build GRCh38
rs753032413 Chr.20: 63698516-63698516 on Build GRCh38
rs777257548 Chr.20: 63698384-63698384 on Build GRCh38
rs767639306 Chr.20: 63698479-63698479 on Build GRCh38
rs759765204 Chr.20: 63698477-63698477 on Build GRCh38
rs768570665 Chr.20: 63698469-63698469 on Build GRCh38
rs776620030 Chr.20: 63698471-63698471 on Build GRCh38
rs757105703 Chr.20: 63698439-63698439 on Build GRCh38
rs756620503 Chr.20: 63698452-63698452 on Build GRCh38
rs771641926 Chr.20: 63698467-63698467 on Build GRCh38
rs778437275 Chr.20: 63698453-63698453 on Build GRCh38
rs774111142 Chr.20: 63698425-63698425 on Build GRCh38
rs575430880 Chr.20: 63698538-63698538 on Build GRCh38
rs749101208 Chr.20: 63698551-63698551 on Build GRCh38
rs747119927 Chr.20: 63698377-63698377 on Build GRCh38
rs550888663 Chr.20: 63698413-63698413 on Build GRCh38
rs771699576 Chr.20: 63698496-63698496 on Build GRCh38
rs538091211 Chr.20: 63698552-63698552 on Build GRCh38
rs767342806 Chr.20: 63698397-63698397 on Build GRCh38
rs747619103 Chr.20: 63698544-63698544 on Build GRCh38
rs768195916 Chr.20: 63698501-63698501 on Build GRCh38
rs764594740 Chr.20: 63698549-63698549 on Build GRCh38
rs202169963 Chr.20: 63698502-63698502 on Build GRCh38
rs748457267 Chr.20: 63698385-63698385 on Build GRCh38
rs779291184 Chr.20: 63698530-63698530 on Build GRCh38
rs764026145 Chr.20: 63698436-63698436 on Build GRCh38
rs753993262 Chr.20: 63698528-63698528 on Build GRCh38
rs770324181 Chr.20: 63698386-63698386 on Build GRCh38
rs372500552 Chr.20: 63698404-63698404 on Build GRCh38
rs751138050 Chr.20: 63698353-63698353 on Build GRCh38
rs746247749 Chr.20: 63698492-63698492 on Build GRCh38

View Full Product Details