Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54181548-54181793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMUG1
Entrez Gene ID
23583
Gene Name
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases
FDG, HMUDG, UNG3
UniGene
Hs.632721
Gene Chromosome Location
Chr.12: 54171815-54188994 on Build GRCh38
Transcripts
NM_001243787.1 NM_001243788.1 NM_001243789.1 NM_001243790.1 NM_001243791.1 NM_014311.2 NR_045039.1 XM_006719319.3 XM_006719320.3 XM_006719321.3 XM_006719322.3 XM_011538109.2 XM_011538110.2 XM_011538111.2 XM_011538112.2 XM_011538113.2 XM_011538114.2 XM_011538115.2 XM_011538116.2 XM_011538117.2 XM_011538118.2 XM_011538119.2 XM_011538120.2 XM_011538121.2 XM_011538122.2 XM_011538123.2 XM_017019113.1 XM_017019114.1 XM_017019115.1 XM_017019116.1 XM_017019117.1 XM_017019118.1 XM_017019119.1 XM_017019120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5361997 c.482T>C p.I161T Chr.12: 54181571-54181571 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs3136394 Chr.12: 54181625-54181625 on Build GRCh38
rs201442554 Chr.12: 54181686-54181686 on Build GRCh38
rs772323337 Chr.12: 54181612-54181612 on Build GRCh38
rs3136392 Chr.12: 54181762-54181762 on Build GRCh38
rs200040688 Chr.12: 54181691-54181691 on Build GRCh38
rs370921169 Chr.12: 54181667-54181667 on Build GRCh38
rs750092921 Chr.12: 54181695-54181695 on Build GRCh38
rs1031962087 Chr.12: 54181645-54181645 on Build GRCh38
rs377626853 Chr.12: 54181597-54181597 on Build GRCh38
rs779627898 Chr.12: 54181594-54181594 on Build GRCh38
rs771205071 Chr.12: 54181631-54181631 on Build GRCh38
rs972239675 Chr.12: 54181711-54181711 on Build GRCh38
rs759303521 Chr.12: 54181601-54181601 on Build GRCh38
rs919447990 Chr.12: 54181742-54181742 on Build GRCh38
rs1032921760 Chr.12: 54181661-54181661 on Build GRCh38
rs774085833 Chr.12: 54181603-54181605 on Build GRCh38
rs971 Chr.12: 54181674-54181674 on Build GRCh38
rs751106121 Chr.12: 54181685-54181685 on Build GRCh38
rs967272177 Chr.12: 54181641-54181641 on Build GRCh38
rs764698993 Chr.12: 54181652-54181652 on Build GRCh38
rs186939603 Chr.12: 54181660-54181660 on Build GRCh38
rs944077113 Chr.12: 54181733-54181733 on Build GRCh38
rs3136393 Chr.12: 54181684-54181684 on Build GRCh38
rs748647964 Chr.12: 54181599-54181599 on Build GRCh38
rs763948835 Chr.12: 54181754-54181754 on Build GRCh38
rs368638453 Chr.12: 54181676-54181676 on Build GRCh38

View Full Product Details