Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132069489-132069736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK26
Entrez Gene ID
51765
Gene Name
serine/threonine protein kinase 26
Gene Aliases
MASK, MST4
UniGene
Hs.444247
Gene Chromosome Location
Chr.X: 132023217-132075943 on Build GRCh38
Transcripts
NM_001042452.1 NM_001042453.1 NM_016542.3 XM_011531349.1 XM_011531350.1

SNP Detail

SNP ID Chromosome Location
rs761165888 Chr.X: 132069552-132069552 on Build GRCh38
rs768152859 Chr.X: 132069541-132069541 on Build GRCh38
rs752950113 Chr.X: 132069693-132069693 on Build GRCh38
rs781583712 Chr.X: 132069545-132069545 on Build GRCh38
rs201682759 Chr.X: 132069593-132069593 on Build GRCh38
rs761137888 Chr.X: 132069643-132069643 on Build GRCh38
rs746225270 Chr.X: 132069683-132069683 on Build GRCh38
rs5933059 Chr.X: 132069692-132069692 on Build GRCh38
rs766997625 Chr.X: 132069576-132069576 on Build GRCh38
rs777970458 Chr.X: 132069712-132069712 on Build GRCh38
rs201758864 Chr.X: 132069528-132069528 on Build GRCh38
rs1033144324 Chr.X: 132069611-132069611 on Build GRCh38
rs778949392 Chr.X: 132069688-132069688 on Build GRCh38
rs762687456 Chr.X: 132069539-132069539 on Build GRCh38
rs376757777 Chr.X: 132069647-132069647 on Build GRCh38
rs754418547 Chr.X: 132069669-132069670 on Build GRCh38
rs750992266 Chr.X: 132069617-132069617 on Build GRCh38
rs759091426 Chr.X: 132069681-132069681 on Build GRCh38
rs754115334 Chr.X: 132069660-132069660 on Build GRCh38
rs200995951 Chr.X: 132069703-132069703 on Build GRCh38

View Full Product Details