Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35535555-35535828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSM2
Entrez Gene ID
84684
Gene Name
INSM transcriptional repressor 2
Gene Aliases
IA-6, IA6, mlt1
UniGene
Hs.62813
Gene Chromosome Location
Chr.14: 35534042-35537054 on Build GRCh38
Transcripts
NM_032594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748824 c.1318T>A p.Y440N Chr.14: 35535570-35535570 on Build GRCh38
5812587 c.1499T>A p.L500Q Chr.14: 35535751-35535751 on Build GRCh38
6521436 c.1555G>A p.G519R Chr.14: 35535807-35535807 on Build GRCh38
4050654 c.1436T>C p.F479S Chr.14: 35535688-35535688 on Build GRCh38
4692663 c.1461G>T p.K487N Chr.14: 35535713-35535713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761773662 Chr.14: 35535589-35535589 on Build GRCh38
rs758768376 Chr.14: 35535695-35535695 on Build GRCh38
rs1007882161 Chr.14: 35535793-35535793 on Build GRCh38
rs771670971 Chr.14: 35535620-35535620 on Build GRCh38
rs765642114 Chr.14: 35535789-35535789 on Build GRCh38
rs760326875 Chr.14: 35535625-35535625 on Build GRCh38
rs768128661 Chr.14: 35535717-35535717 on Build GRCh38
rs774808114 Chr.14: 35535622-35535622 on Build GRCh38
rs764266222 Chr.14: 35535782-35535782 on Build GRCh38
rs373080462 Chr.14: 35535612-35535612 on Build GRCh38
rs545517333 Chr.14: 35535790-35535790 on Build GRCh38
rs763893619 Chr.14: 35535577-35535577 on Build GRCh38
rs777661164 Chr.14: 35535720-35535721 on Build GRCh38
rs753445756 Chr.14: 35535792-35535793 on Build GRCh38
rs201878584 Chr.14: 35535796-35535796 on Build GRCh38
rs766186463 Chr.14: 35535771-35535771 on Build GRCh38
rs142314509 Chr.14: 35535598-35535598 on Build GRCh38
rs866863375 Chr.14: 35535806-35535806 on Build GRCh38
rs776302314 Chr.14: 35535735-35535735 on Build GRCh38
rs753101993 Chr.14: 35535704-35535704 on Build GRCh38
rs1014247924 Chr.14: 35535624-35535624 on Build GRCh38
rs879211045 Chr.14: 35535630-35535630 on Build GRCh38
rs752733436 Chr.14: 35535630-35535631 on Build GRCh38
rs755121149 Chr.14: 35535606-35535606 on Build GRCh38
rs151277868 Chr.14: 35535616-35535616 on Build GRCh38
rs754173185 Chr.14: 35535784-35535784 on Build GRCh38
rs369209579 Chr.14: 35535608-35535608 on Build GRCh38
rs765169463 Chr.14: 35535596-35535596 on Build GRCh38
rs756436213 Chr.14: 35535662-35535662 on Build GRCh38
rs759478927 Chr.14: 35535607-35535608 on Build GRCh38
rs965914054 Chr.14: 35535773-35535773 on Build GRCh38
rs200612952 Chr.14: 35535794-35535794 on Build GRCh38
rs140515178 Chr.14: 35535628-35535628 on Build GRCh38
rs376053074 Chr.14: 35535617-35535617 on Build GRCh38
rs768261524 Chr.14: 35535626-35535626 on Build GRCh38
rs754083126 Chr.14: 35535671-35535671 on Build GRCh38
rs536480490 Chr.14: 35535685-35535685 on Build GRCh38
rs769685229 Chr.14: 35535746-35535746 on Build GRCh38
rs186564978 Chr.14: 35535716-35535716 on Build GRCh38
rs1014890945 Chr.14: 35535756-35535756 on Build GRCh38
rs756616256 Chr.14: 35535769-35535769 on Build GRCh38
rs190091421 Chr.14: 35535807-35535807 on Build GRCh38
rs750302555 Chr.14: 35535601-35535601 on Build GRCh38
rs778091935 Chr.14: 35535669-35535669 on Build GRCh38
rs753566423 Chr.14: 35535587-35535587 on Build GRCh38
rs369133835 Chr.14: 35535644-35535644 on Build GRCh38
rs75674743 Chr.14: 35535723-35535723 on Build GRCh38
rs772824179 Chr.14: 35535750-35535750 on Build GRCh38
rs762951462 Chr.14: 35535765-35535765 on Build GRCh38
rs750438881 Chr.14: 35535633-35535633 on Build GRCh38
rs746401759 Chr.14: 35535693-35535693 on Build GRCh38
rs752756480 Chr.14: 35535657-35535657 on Build GRCh38
rs1005117633 Chr.14: 35535745-35535745 on Build GRCh38
rs755839603 Chr.14: 35535705-35535705 on Build GRCh38
rs762783877 Chr.14: 35535643-35535643 on Build GRCh38
rs576651153 Chr.14: 35535741-35535741 on Build GRCh38
rs1045667593 Chr.14: 35535658-35535658 on Build GRCh38
rs779133987 Chr.14: 35535690-35535690 on Build GRCh38
rs1001595023 Chr.14: 35535696-35535696 on Build GRCh38
rs554053535 Chr.14: 35535613-35535613 on Build GRCh38
rs1052133700 Chr.14: 35535691-35535691 on Build GRCh38
rs779649832 Chr.14: 35535702-35535702 on Build GRCh38
rs767352675 Chr.14: 35535627-35535628 on Build GRCh38
rs756346517 Chr.14: 35535611-35535611 on Build GRCh38
rs764906496 Chr.14: 35535631-35535631 on Build GRCh38
rs760879378 Chr.14: 35535772-35535772 on Build GRCh38

View Full Product Details