Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66429186-66429449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRS1
Entrez Gene ID
23212
Gene Name
ribosome biogenesis regulator homolog
Gene Aliases
-
UniGene
Hs.71827
Gene Chromosome Location
Chr.8: 66429028-66430733 on Build GRCh38
Transcripts
NM_015169.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6239375 c.145G>C p.G49R Chr.8: 66429276-66429276 on Build GRCh38
4922358 c.117G>A p.L39L Chr.8: 66429248-66429248 on Build GRCh38
6342897 c.99G>T p.Q33H Chr.8: 66429230-66429230 on Build GRCh38
5800438 c.187G>A p.A63T Chr.8: 66429318-66429318 on Build GRCh38
1101059 c.111C>T p.G37G Chr.8: 66429242-66429242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964388256 Chr.8: 66429414-66429414 on Build GRCh38
rs536702091 Chr.8: 66429266-66429266 on Build GRCh38
rs867283216 Chr.8: 66429239-66429239 on Build GRCh38
rs907465920 Chr.8: 66429305-66429305 on Build GRCh38
rs749093852 Chr.8: 66429207-66429207 on Build GRCh38
rs959600407 Chr.8: 66429423-66429423 on Build GRCh38
rs751244628 Chr.8: 66429258-66429258 on Build GRCh38
rs556647477 Chr.8: 66429389-66429389 on Build GRCh38
rs749044838 Chr.8: 66429271-66429271 on Build GRCh38
rs777583115 Chr.8: 66429267-66429267 on Build GRCh38
rs116590536 Chr.8: 66429246-66429246 on Build GRCh38
rs974396127 Chr.8: 66429431-66429431 on Build GRCh38
rs867587737 Chr.8: 66429240-66429240 on Build GRCh38
rs761755888 Chr.8: 66429348-66429348 on Build GRCh38
rs775859459 Chr.8: 66429215-66429215 on Build GRCh38
rs774747447 Chr.8: 66429316-66429316 on Build GRCh38
rs11556131 Chr.8: 66429309-66429309 on Build GRCh38
rs773601050 Chr.8: 66429299-66429299 on Build GRCh38
rs777595652 Chr.8: 66429206-66429206 on Build GRCh38
rs770280890 Chr.8: 66429298-66429298 on Build GRCh38
rs1021380760 Chr.8: 66429397-66429397 on Build GRCh38
rs780371153 Chr.8: 66429276-66429276 on Build GRCh38
rs200355092 Chr.8: 66429221-66429221 on Build GRCh38
rs367878265 Chr.8: 66429280-66429280 on Build GRCh38
rs371751925 Chr.8: 66429314-66429314 on Build GRCh38
rs769038085 Chr.8: 66429285-66429285 on Build GRCh38
rs776764649 Chr.8: 66429290-66429290 on Build GRCh38
rs970465469 Chr.8: 66429382-66429382 on Build GRCh38
rs748238240 Chr.8: 66429295-66429295 on Build GRCh38
rs148041062 Chr.8: 66429257-66429257 on Build GRCh38
rs763340999 Chr.8: 66429308-66429308 on Build GRCh38
rs997079184 Chr.8: 66429278-66429278 on Build GRCh38
rs375178198 Chr.8: 66429216-66429216 on Build GRCh38
rs890562662 Chr.8: 66429260-66429260 on Build GRCh38
rs576594281 Chr.8: 66429405-66429405 on Build GRCh38
rs767226444 Chr.8: 66429380-66429380 on Build GRCh38
rs757149111 Chr.8: 66429273-66429273 on Build GRCh38
rs753741684 Chr.8: 66429424-66429424 on Build GRCh38
rs1003908775 Chr.8: 66429328-66429328 on Build GRCh38
rs1040905517 Chr.8: 66429275-66429275 on Build GRCh38
rs759159589 Chr.8: 66429354-66429354 on Build GRCh38
rs1032280451 Chr.8: 66429334-66429334 on Build GRCh38
rs868431619 Chr.8: 66429369-66429369 on Build GRCh38
rs748622273 Chr.8: 66429265-66429266 on Build GRCh38
rs755582456 Chr.8: 66429396-66429396 on Build GRCh38
rs765734494 Chr.8: 66429229-66429229 on Build GRCh38
rs140840609 Chr.8: 66429261-66429261 on Build GRCh38
rs762347986 Chr.8: 66429226-66429226 on Build GRCh38

View Full Product Details