Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69161469-69161708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PJA1
Entrez Gene ID
64219
Gene Name
praja ring finger ubiquitin ligase 1
Gene Aliases
PRAJA1, RNF70
UniGene
Hs.522679
Gene Chromosome Location
Chr.X: 69160738-69165522 on Build GRCh38
Transcripts
NM_001032396.2 NM_022368.4 NM_145119.3 XM_005262292.1 XM_011531011.2

SNP Detail

SNP ID Chromosome Location
rs777932711 Chr.X: 69161496-69161496 on Build GRCh38
rs766084279 Chr.X: 69161555-69161555 on Build GRCh38
rs190011418 Chr.X: 69161547-69161547 on Build GRCh38
rs759369667 Chr.X: 69161570-69161570 on Build GRCh38
rs758410215 Chr.X: 69161619-69161619 on Build GRCh38
rs746327146 Chr.X: 69161518-69161518 on Build GRCh38
rs776158176 Chr.X: 69161528-69161528 on Build GRCh38
rs770378843 Chr.X: 69161521-69161521 on Build GRCh38
rs867864585 Chr.X: 69161504-69161504 on Build GRCh38
rs776530941 Chr.X: 69161565-69161565 on Build GRCh38
rs760431439 Chr.X: 69161553-69161553 on Build GRCh38
rs200830501 Chr.X: 69161513-69161513 on Build GRCh38
rs1046448562 Chr.X: 69161512-69161512 on Build GRCh38
rs747621479 Chr.X: 69161538-69161538 on Build GRCh38
rs201523833 Chr.X: 69161542-69161542 on Build GRCh38
rs1033820486 Chr.X: 69161660-69161660 on Build GRCh38
rs199707327 Chr.X: 69161581-69161581 on Build GRCh38
rs772939881 Chr.X: 69161551-69161551 on Build GRCh38
rs199869943 Chr.X: 69161493-69161493 on Build GRCh38
rs757508919 Chr.X: 69161506-69161506 on Build GRCh38
rs747239855 Chr.X: 69161505-69161505 on Build GRCh38
rs146361461 Chr.X: 69161580-69161580 on Build GRCh38
rs764175826 Chr.X: 69161631-69161631 on Build GRCh38

View Full Product Details