Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 53809020-53809249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH15
Entrez Gene ID
65217
Gene Name
protocadherin related 15
Gene Aliases
CDHR15, DFNB23, USH1F
UniGene
Hs.280209
Gene Chromosome Location
Chr.10: 53802771-55629182 on Build GRCh38
Transcripts
NM_001142769.1 NM_001142770.1 NM_001142771.1 NM_001142772.1 XM_017016571.1 XM_017016572.1 XM_017016573.1

SNP Detail

SNP ID Chromosome Location
rs921529479 Chr.10: 53809168-53809168 on Build GRCh38
rs976033642 Chr.10: 53809145-53809145 on Build GRCh38
rs730880021 Chr.10: 53809138-53809138 on Build GRCh38
rs755469136 Chr.10: 53809115-53809115 on Build GRCh38
rs746920651 Chr.10: 53809078-53809078 on Build GRCh38
rs745608643 Chr.10: 53809053-53809053 on Build GRCh38
rs778583665 Chr.10: 53809052-53809052 on Build GRCh38
rs772876205 Chr.10: 53809174-53809174 on Build GRCh38
rs534167845 Chr.10: 53809148-53809148 on Build GRCh38
rs867467512 Chr.10: 53809082-53809082 on Build GRCh38
rs531574437 Chr.10: 53809211-53809211 on Build GRCh38
rs776703713 Chr.10: 53809109-53809109 on Build GRCh38
rs765728490 Chr.10: 53809088-53809088 on Build GRCh38
rs369936199 Chr.10: 53809150-53809150 on Build GRCh38
rs778088114 Chr.10: 53809136-53809136 on Build GRCh38
rs747630384 Chr.10: 53809161-53809161 on Build GRCh38
rs768321475 Chr.10: 53809080-53809080 on Build GRCh38
rs776020523 Chr.10: 53809160-53809160 on Build GRCh38
rs762576591 Chr.10: 53809178-53809178 on Build GRCh38
rs866152374 Chr.10: 53809069-53809069 on Build GRCh38
rs754203301 Chr.10: 53809055-53809055 on Build GRCh38
rs776201814 Chr.10: 53809087-53809087 on Build GRCh38
rs774682018 Chr.10: 53809185-53809185 on Build GRCh38
rs544980556 Chr.10: 53809092-53809092 on Build GRCh38
rs758316594 Chr.10: 53809146-53809146 on Build GRCh38
rs181269166 Chr.10: 53809134-53809134 on Build GRCh38
rs769366626 Chr.10: 53809162-53809162 on Build GRCh38
rs745671714 Chr.10: 53809137-53809137 on Build GRCh38
rs752753497 Chr.10: 53809098-53809099 on Build GRCh38
rs767998740 Chr.10: 53809196-53809196 on Build GRCh38
rs747270113 Chr.10: 53809153-53809154 on Build GRCh38
rs752114183 Chr.10: 53809105-53809105 on Build GRCh38
rs766793106 Chr.10: 53809184-53809184 on Build GRCh38
rs369664517 Chr.10: 53809221-53809221 on Build GRCh38
rs779964014 Chr.10: 53809149-53809149 on Build GRCh38
rs183631592 Chr.10: 53809116-53809116 on Build GRCh38
rs755495232 Chr.10: 53809056-53809058 on Build GRCh38
rs772165950 Chr.10: 53809073-53809073 on Build GRCh38
rs760046709 Chr.10: 53809187-53809187 on Build GRCh38
rs758562162 Chr.10: 53809106-53809107 on Build GRCh38
rs372184022 Chr.10: 53809201-53809201 on Build GRCh38
rs763267318 Chr.10: 53809097-53809097 on Build GRCh38
rs756168953 Chr.10: 53809205-53809205 on Build GRCh38
rs756342988 Chr.10: 53809049-53809049 on Build GRCh38
rs373988170 Chr.10: 53809102-53809102 on Build GRCh38
rs761431484 Chr.10: 53809088-53809088 on Build GRCh38
rs775518290 Chr.10: 53809076-53809076 on Build GRCh38
rs11003863 Chr.10: 53809212-53809212 on Build GRCh38
rs16937768 Chr.10: 53809118-53809118 on Build GRCh38
rs757596933 Chr.10: 53809163-53809164 on Build GRCh38

View Full Product Details