Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23199389-23199636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUPL2
Entrez Gene ID
11097
Gene Name
nucleoporin like 2
Gene Aliases
CG1, NLP-1, NLP_1, hCG1
UniGene
Hs.408241
Gene Chromosome Location
Chr.7: 23181567-23201011 on Build GRCh38
Transcripts
NM_007342.2 XM_005249592.1 XM_005249593.3 XM_011515104.2 XM_017011714.1 XM_017011715.1 XM_017011716.1 XM_017011717.1 XM_017011718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3880261 c.690G>A p.S230S Chr.7: 23199538-23199538 on Build GRCh38
3637510 c.645A>T p.A215A Chr.7: 23199493-23199493 on Build GRCh38
4587327 c.689C>T p.S230L Chr.7: 23199537-23199537 on Build GRCh38
4155699 c.694+7G>T p.? Chr.7: 23199549-23199549 on Build GRCh38
22097 c.647C>T p.P216L Chr.7: 23199495-23199495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746108294 Chr.7: 23199431-23199431 on Build GRCh38
rs201901418 Chr.7: 23199537-23199537 on Build GRCh38
rs569259219 Chr.7: 23199461-23199461 on Build GRCh38
rs766197271 Chr.7: 23199494-23199494 on Build GRCh38
rs551214272 Chr.7: 23199445-23199445 on Build GRCh38
rs764386269 Chr.7: 23199578-23199578 on Build GRCh38
rs771507036 Chr.7: 23199465-23199465 on Build GRCh38
rs201316346 Chr.7: 23199456-23199456 on Build GRCh38
rs369615940 Chr.7: 23199444-23199445 on Build GRCh38
rs535663890 Chr.7: 23199600-23199600 on Build GRCh38
rs763366673 Chr.7: 23199577-23199577 on Build GRCh38
rs1035611145 Chr.7: 23199420-23199420 on Build GRCh38
rs1016044835 Chr.7: 23199427-23199427 on Build GRCh38
rs140947183 Chr.7: 23199529-23199529 on Build GRCh38
rs774690348 Chr.7: 23199483-23199488 on Build GRCh38
rs371504782 Chr.7: 23199521-23199521 on Build GRCh38
rs1057082452 Chr.7: 23199560-23199560 on Build GRCh38
rs774197609 Chr.7: 23199460-23199460 on Build GRCh38
rs763787396 Chr.7: 23199445-23199446 on Build GRCh38
rs961647625 Chr.7: 23199438-23199438 on Build GRCh38
rs200616527 Chr.7: 23199520-23199520 on Build GRCh38
rs764628696 Chr.7: 23199513-23199513 on Build GRCh38
rs772868016 Chr.7: 23199472-23199472 on Build GRCh38
rs910306430 Chr.7: 23199533-23199533 on Build GRCh38
rs188931541 Chr.7: 23199559-23199559 on Build GRCh38
rs780303424 Chr.7: 23199538-23199538 on Build GRCh38
rs757310818 Chr.7: 23199517-23199517 on Build GRCh38
rs746571277 Chr.7: 23199564-23199564 on Build GRCh38
rs185054777 Chr.7: 23199550-23199550 on Build GRCh38
rs858237 Chr.7: 23199604-23199604 on Build GRCh38
rs770680270 Chr.7: 23199571-23199571 on Build GRCh38
rs924023238 Chr.7: 23199523-23199523 on Build GRCh38
rs762230684 Chr.7: 23199588-23199588 on Build GRCh38
rs768558485 Chr.7: 23199457-23199457 on Build GRCh38
rs754761597 Chr.7: 23199540-23199540 on Build GRCh38
rs200016707 Chr.7: 23199528-23199528 on Build GRCh38
rs748052269 Chr.7: 23199553-23199553 on Build GRCh38
rs781434474 Chr.7: 23199425-23199425 on Build GRCh38
rs763457240 Chr.7: 23199496-23199496 on Build GRCh38
rs769520905 Chr.7: 23199443-23199443 on Build GRCh38
rs771548143 Chr.7: 23199557-23199557 on Build GRCh38
rs371751501 Chr.7: 23199581-23199581 on Build GRCh38
rs858238 Chr.7: 23199514-23199514 on Build GRCh38
rs779828367 Chr.7: 23199452-23199452 on Build GRCh38
rs998418027 Chr.7: 23199466-23199466 on Build GRCh38
rs760411677 Chr.7: 23199476-23199476 on Build GRCh38
rs978445193 Chr.7: 23199539-23199539 on Build GRCh38
rs1029772208 Chr.7: 23199492-23199492 on Build GRCh38
rs368811281 Chr.7: 23199575-23199575 on Build GRCh38
rs775980109 Chr.7: 23199495-23199495 on Build GRCh38
rs917062951 Chr.7: 23199596-23199596 on Build GRCh38
rs1022587610 Chr.7: 23199490-23199490 on Build GRCh38

View Full Product Details