Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197917850-197918111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX9
Entrez Gene ID
56956
Gene Name
LIM homeobox 9
Gene Aliases
-
UniGene
Hs.442578
Gene Chromosome Location
Chr.1: 197912505-197935476 on Build GRCh38
Transcripts
NM_001014434.1 NM_020204.2 XM_005245350.3 XM_011509781.2 XM_017001849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400167 c.108C>T p.A36A Chr.1: 197917958-197917958 on Build GRCh38
2125764 c.36G>A p.S12S Chr.1: 197917859-197917859 on Build GRCh38
4614689 c.48delC p.P18fs*76 Chr.1: 197917871-197917871 on Build GRCh38
4557461 c.72G>A p.G24G Chr.1: 197917895-197917895 on Build GRCh38
301274 c.33G>C p.M11I Chr.1: 197917883-197917883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776528891 Chr.1: 197917934-197917934 on Build GRCh38
rs920489029 Chr.1: 197917960-197917960 on Build GRCh38
rs746885535 Chr.1: 197918015-197918015 on Build GRCh38
rs374479359 Chr.1: 197917961-197917961 on Build GRCh38
rs777583081 Chr.1: 197917925-197917925 on Build GRCh38
rs144348823 Chr.1: 197917990-197917990 on Build GRCh38
rs974075181 Chr.1: 197917951-197917951 on Build GRCh38
rs769590567 Chr.1: 197917944-197917944 on Build GRCh38
rs762208252 Chr.1: 197918036-197918036 on Build GRCh38
rs756372594 Chr.1: 197917896-197917896 on Build GRCh38
rs147454219 Chr.1: 197917975-197917975 on Build GRCh38
rs781663927 Chr.1: 197917883-197917883 on Build GRCh38
rs780087696 Chr.1: 197917901-197917901 on Build GRCh38
rs12091190 Chr.1: 197917878-197917878 on Build GRCh38
rs544478215 Chr.1: 197918091-197918091 on Build GRCh38
rs767745554 Chr.1: 197917974-197917974 on Build GRCh38
rs1045476161 Chr.1: 197918071-197918071 on Build GRCh38
rs749395451 Chr.1: 197917907-197917907 on Build GRCh38
rs201760984 Chr.1: 197917922-197917922 on Build GRCh38
rs747021625 Chr.1: 197917927-197917927 on Build GRCh38
rs577856015 Chr.1: 197917972-197917972 on Build GRCh38
rs573388724 Chr.1: 197917994-197917994 on Build GRCh38
rs113693840 Chr.1: 197917965-197917965 on Build GRCh38
rs542681982 Chr.1: 197917957-197917957 on Build GRCh38
rs780288990 Chr.1: 197917985-197917985 on Build GRCh38
rs562140354 Chr.1: 197918039-197918039 on Build GRCh38
rs745687088 Chr.1: 197918023-197918023 on Build GRCh38
rs756282716 Chr.1: 197917980-197917980 on Build GRCh38
rs902947386 Chr.1: 197918074-197918074 on Build GRCh38
rs773955143 Chr.1: 197918032-197918032 on Build GRCh38
rs761038021 Chr.1: 197918041-197918041 on Build GRCh38
rs759241758 Chr.1: 197917939-197917939 on Build GRCh38
rs148027167 Chr.1: 197917871-197917871 on Build GRCh38
rs755035631 Chr.1: 197917991-197917991 on Build GRCh38
rs16842109 Chr.1: 197918057-197918057 on Build GRCh38
rs893268142 Chr.1: 197917898-197917898 on Build GRCh38
rs770623864 Chr.1: 197917930-197917930 on Build GRCh38
rs201746672 Chr.1: 197917973-197917973 on Build GRCh38
rs762754035 Chr.1: 197918030-197918030 on Build GRCh38
rs757576735 Chr.1: 197917875-197917875 on Build GRCh38
rs775297308 Chr.1: 197918029-197918029 on Build GRCh38
rs200517180 Chr.1: 197918019-197918019 on Build GRCh38
rs780800892 Chr.1: 197918021-197918021 on Build GRCh38
rs771927350 Chr.1: 197917913-197917913 on Build GRCh38
rs764606159 Chr.1: 197917971-197917971 on Build GRCh38
rs769502409 Chr.1: 197918027-197918027 on Build GRCh38
rs990942306 Chr.1: 197918077-197918077 on Build GRCh38
rs1029168176 Chr.1: 197917873-197917873 on Build GRCh38
rs746077703 Chr.1: 197917893-197917893 on Build GRCh38
rs960502139 Chr.1: 197918072-197918072 on Build GRCh38
rs916418116 Chr.1: 197918090-197918090 on Build GRCh38

View Full Product Details