Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175937317-175937558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LNPK
Entrez Gene ID
80856
Gene Name
lunapark, ER junction formation factor
Gene Aliases
KIAA1715, LNP, LNP1, Ul, ulnaless
UniGene
Hs.209561
Gene Chromosome Location
Chr.2: 175923862-176002901 on Build GRCh38
Transcripts
NM_001305008.1 NM_001305009.1 NM_001305010.1 NM_001305011.1 NM_030650.2 NR_130941.1 XM_005246878.3 XM_005246879.4 XM_005246880.3 XM_006712783.2 XM_011511945.2 XM_017005055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295750 c.1135C>A p.Q379K Chr.2: 175937356-175937356 on Build GRCh38
5952227 c.1070A>T p.Q357L Chr.2: 175937421-175937421 on Build GRCh38
4812788 c.977C>G p.A326G Chr.2: 175937514-175937514 on Build GRCh38
1401408 c.900C>A p.Y300* Chr.2: 175937498-175937498 on Build GRCh38
4875675 c.1016G>T p.R339I Chr.2: 175937475-175937475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008832394 Chr.2: 175937479-175937479 on Build GRCh38
rs1040316125 Chr.2: 175937397-175937397 on Build GRCh38
rs554332202 Chr.2: 175937510-175937510 on Build GRCh38
rs1047835794 Chr.2: 175937503-175937503 on Build GRCh38
rs751039929 Chr.2: 175937520-175937520 on Build GRCh38
rs200017613 Chr.2: 175937418-175937418 on Build GRCh38
rs568552587 Chr.2: 175937459-175937459 on Build GRCh38
rs568599024 Chr.2: 175937524-175937524 on Build GRCh38
rs764249318 Chr.2: 175937506-175937506 on Build GRCh38
rs762785151 Chr.2: 175937358-175937358 on Build GRCh38
rs556625914 Chr.2: 175937423-175937423 on Build GRCh38
rs764316272 Chr.2: 175937384-175937384 on Build GRCh38
rs753864965 Chr.2: 175937385-175937385 on Build GRCh38
rs201325808 Chr.2: 175937392-175937392 on Build GRCh38
rs754039320 Chr.2: 175937509-175937509 on Build GRCh38
rs1029004757 Chr.2: 175937429-175937429 on Build GRCh38
rs780831858 Chr.2: 175937420-175937420 on Build GRCh38
rs537849084 Chr.2: 175937417-175937417 on Build GRCh38
rs746360310 Chr.2: 175937394-175937394 on Build GRCh38
rs762871560 Chr.2: 175937474-175937474 on Build GRCh38
rs752982922 Chr.2: 175937381-175937381 on Build GRCh38
rs759257720 Chr.2: 175937502-175937502 on Build GRCh38
rs147174572 Chr.2: 175937501-175937501 on Build GRCh38
rs758877994 Chr.2: 175937409-175937409 on Build GRCh38
rs901853053 Chr.2: 175937398-175937398 on Build GRCh38
rs757317200 Chr.2: 175937390-175937390 on Build GRCh38
rs1000004733 Chr.2: 175937377-175937377 on Build GRCh38
rs780565202 Chr.2: 175937416-175937416 on Build GRCh38
rs1010947172 Chr.2: 175937427-175937427 on Build GRCh38
rs765332981 Chr.2: 175937518-175937518 on Build GRCh38
rs766130188 Chr.2: 175937370-175937370 on Build GRCh38
rs139043546 Chr.2: 175937460-175937460 on Build GRCh38
rs770768520 Chr.2: 175937499-175937499 on Build GRCh38
rs200237452 Chr.2: 175937522-175937522 on Build GRCh38

View Full Product Details