Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67935890-67936123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB10
Entrez Gene ID
5699
Gene Name
proteasome subunit beta 10
Gene Aliases
LMP10, MECL1, beta2i
UniGene
Hs.9661
Gene Chromosome Location
Chr.16: 67934504-67936877 on Build GRCh38
Transcripts
NM_002801.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4718432 c.312A>G p.L104L Chr.16: 67936034-67936034 on Build GRCh38
3511145 c.348C>T p.A116A Chr.16: 67935998-67935998 on Build GRCh38
5851267 c.337C>T p.P113S Chr.16: 67936009-67936009 on Build GRCh38
3511146 c.323C>A p.S108Y Chr.16: 67936023-67936023 on Build GRCh38
3754987 c.319T>C p.L107L Chr.16: 67936027-67936027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754244406 Chr.16: 67935935-67935935 on Build GRCh38
rs764556949 Chr.16: 67936032-67936032 on Build GRCh38
rs758135754 Chr.16: 67936086-67936086 on Build GRCh38
rs990613016 Chr.16: 67936061-67936061 on Build GRCh38
rs750554058 Chr.16: 67936063-67936063 on Build GRCh38
rs777849619 Chr.16: 67935914-67935914 on Build GRCh38
rs753155097 Chr.16: 67935954-67935954 on Build GRCh38
rs1050654704 Chr.16: 67935963-67935963 on Build GRCh38
rs770335398 Chr.16: 67935988-67935988 on Build GRCh38
rs777607072 Chr.16: 67936095-67936095 on Build GRCh38
rs778538577 Chr.16: 67935974-67935974 on Build GRCh38
rs535762295 Chr.16: 67936067-67936067 on Build GRCh38
rs1001085577 Chr.16: 67935995-67935995 on Build GRCh38
rs570917522 Chr.16: 67935962-67935962 on Build GRCh38
rs765922650 Chr.16: 67935933-67935933 on Build GRCh38
rs760140635 Chr.16: 67935931-67935931 on Build GRCh38
rs753711137 Chr.16: 67936098-67936099 on Build GRCh38
rs771888401 Chr.16: 67935979-67935979 on Build GRCh38
rs370963927 Chr.16: 67935956-67935956 on Build GRCh38
rs778872378 Chr.16: 67936070-67936070 on Build GRCh38
rs763368478 Chr.16: 67936028-67936028 on Build GRCh38
rs201566321 Chr.16: 67935916-67935916 on Build GRCh38
rs749163355 Chr.16: 67935966-67935966 on Build GRCh38
rs760147515 Chr.16: 67935938-67935938 on Build GRCh38
rs200453196 Chr.16: 67935958-67935958 on Build GRCh38
rs772788981 Chr.16: 67935980-67935980 on Build GRCh38
rs764094117 Chr.16: 67936017-67936017 on Build GRCh38
rs768702177 Chr.16: 67935968-67935968 on Build GRCh38
rs923044203 Chr.16: 67935927-67935927 on Build GRCh38
rs757553185 Chr.16: 67936044-67936044 on Build GRCh38
rs781342387 Chr.16: 67935965-67935965 on Build GRCh38
rs771749532 Chr.16: 67935925-67935925 on Build GRCh38
rs765515451 Chr.16: 67935940-67935940 on Build GRCh38
rs202238433 Chr.16: 67936012-67936012 on Build GRCh38
rs767485954 Chr.16: 67936050-67936050 on Build GRCh38
rs751773840 Chr.16: 67936041-67936041 on Build GRCh38
rs747892028 Chr.16: 67935977-67935977 on Build GRCh38
rs1023473539 Chr.16: 67936024-67936024 on Build GRCh38
rs755525249 Chr.16: 67935921-67935922 on Build GRCh38
rs20549 Chr.16: 67936027-67936027 on Build GRCh38
rs748084838 Chr.16: 67936081-67936081 on Build GRCh38
rs772876833 Chr.16: 67935930-67935930 on Build GRCh38
rs765823898 Chr.16: 67936019-67936019 on Build GRCh38
rs200059670 Chr.16: 67935985-67935985 on Build GRCh38
rs148173943 Chr.16: 67935996-67935996 on Build GRCh38
rs1013570966 Chr.16: 67936030-67936030 on Build GRCh38

View Full Product Details