Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47187244-47187493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZK1IP1
Entrez Gene ID
10158
Gene Name
PDZK1 interacting protein 1
Gene Aliases
DD96, MAP17, SPAP
UniGene
Hs.431099
Gene Chromosome Location
Chr.1: 47183589-47190099 on Build GRCh38
Transcripts
NM_005764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7383503 c.106G>A p.A36T Chr.1: 47187389-47187389 on Build GRCh38
7282925 c.176C>T p.P59L Chr.1: 47187319-47187319 on Build GRCh38
3865718 c.153C>T p.N51N Chr.1: 47187342-47187342 on Build GRCh38
1343202 c.130G>A p.V44I Chr.1: 47187365-47187365 on Build GRCh38
7252721 c.147A>T p.A49A Chr.1: 47187348-47187348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548754046 Chr.1: 47187315-47187315 on Build GRCh38
rs1024248149 Chr.1: 47187462-47187462 on Build GRCh38
rs373545384 Chr.1: 47187271-47187271 on Build GRCh38
rs775178511 Chr.1: 47187418-47187418 on Build GRCh38
rs760959554 Chr.1: 47187275-47187275 on Build GRCh38
rs77207461 Chr.1: 47187387-47187387 on Build GRCh38
rs776952702 Chr.1: 47187419-47187419 on Build GRCh38
rs953182462 Chr.1: 47187392-47187392 on Build GRCh38
rs986684539 Chr.1: 47187406-47187406 on Build GRCh38
rs780551627 Chr.1: 47187368-47187368 on Build GRCh38
rs774226990 Chr.1: 47187319-47187319 on Build GRCh38
rs756259974 Chr.1: 47187373-47187373 on Build GRCh38
rs768279277 Chr.1: 47187340-47187340 on Build GRCh38
rs200901884 Chr.1: 47187417-47187417 on Build GRCh38
rs779603665 Chr.1: 47187288-47187288 on Build GRCh38
rs780756510 Chr.1: 47187301-47187301 on Build GRCh38
rs749875677 Chr.1: 47187468-47187468 on Build GRCh38
rs749864358 Chr.1: 47187312-47187315 on Build GRCh38
rs767610762 Chr.1: 47187268-47187268 on Build GRCh38
rs1051090112 Chr.1: 47187348-47187348 on Build GRCh38
rs11552561 Chr.1: 47187365-47187365 on Build GRCh38
rs369958859 Chr.1: 47187389-47187389 on Build GRCh38
rs773162789 Chr.1: 47187405-47187405 on Build GRCh38
rs200717593 Chr.1: 47187388-47187388 on Build GRCh38
rs772106467 Chr.1: 47187396-47187396 on Build GRCh38
rs749117898 Chr.1: 47187314-47187314 on Build GRCh38
rs759841391 Chr.1: 47187435-47187435 on Build GRCh38
rs779800928 Chr.1: 47187381-47187381 on Build GRCh38
rs756786145 Chr.1: 47187298-47187298 on Build GRCh38
rs773443753 Chr.1: 47187297-47187297 on Build GRCh38
rs182360315 Chr.1: 47187357-47187357 on Build GRCh38
rs754121679 Chr.1: 47187346-47187346 on Build GRCh38
rs201358886 Chr.1: 47187316-47187316 on Build GRCh38
rs771318647 Chr.1: 47187316-47187317 on Build GRCh38
rs755538552 Chr.1: 47187284-47187284 on Build GRCh38
rs183333562 Chr.1: 47187476-47187476 on Build GRCh38
rs17850898 Chr.1: 47187342-47187342 on Build GRCh38
rs748965644 Chr.1: 47187382-47187382 on Build GRCh38
rs773358087 Chr.1: 47187329-47187329 on Build GRCh38
rs776053687 Chr.1: 47187456-47187456 on Build GRCh38
rs35269328 Chr.1: 47187408-47187408 on Build GRCh38
rs766597470 Chr.1: 47187279-47187279 on Build GRCh38
rs200168915 Chr.1: 47187380-47187380 on Build GRCh38
rs958266753 Chr.1: 47187451-47187451 on Build GRCh38
rs150502884 Chr.1: 47187318-47187318 on Build GRCh38
rs753278067 Chr.1: 47187291-47187291 on Build GRCh38
rs755597813 Chr.1: 47187471-47187471 on Build GRCh38
rs771036728 Chr.1: 47187413-47187413 on Build GRCh38
rs775058429 Chr.1: 47187366-47187366 on Build GRCh38
rs368888060 Chr.1: 47187464-47187464 on Build GRCh38
rs369843724 Chr.1: 47187302-47187302 on Build GRCh38
rs779705538 Chr.1: 47187312-47187312 on Build GRCh38
rs376997785 Chr.1: 47187448-47187448 on Build GRCh38
rs754288621 Chr.1: 47187282-47187282 on Build GRCh38
rs760797102 Chr.1: 47187339-47187339 on Build GRCh38
rs759931017 Chr.1: 47187356-47187356 on Build GRCh38
rs763547123 Chr.1: 47187457-47187457 on Build GRCh38
rs769541904 Chr.1: 47187309-47187309 on Build GRCh38
rs201952169 Chr.1: 47187320-47187320 on Build GRCh38
rs199783464 Chr.1: 47187294-47187294 on Build GRCh38
rs372002988 Chr.1: 47187327-47187327 on Build GRCh38

View Full Product Details