Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5514835-5515100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP1
Entrez Gene ID
22861
Gene Name
NLR family pyrin domain containing 1
Gene Aliases
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1
UniGene
Hs.652273
Gene Chromosome Location
Chr.17: 5501399-5584512 on Build GRCh38
Transcripts
NM_001033053.2 NM_014922.4 NM_033004.3 NM_033006.3 NM_033007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5018416 c.4269delC p.S1424fs*9 Chr.17: 5514907-5514907 on Build GRCh38
6886834 c.4311C>T p.D1437D Chr.17: 5514865-5514865 on Build GRCh38
4384880 c.4219G>T p.V1407L Chr.17: 5514957-5514957 on Build GRCh38
6727026 c.4227C>T p.S1409S Chr.17: 5514949-5514949 on Build GRCh38
6457586 c.4313G>A p.R1438Q Chr.17: 5514863-5514863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775960305 Chr.17: 5515065-5515065 on Build GRCh38
rs925924700 Chr.17: 5515059-5515059 on Build GRCh38
rs202117967 Chr.17: 5515072-5515072 on Build GRCh38
rs368481005 Chr.17: 5514910-5514910 on Build GRCh38
rs748877010 Chr.17: 5514936-5514936 on Build GRCh38
rs748024457 Chr.17: 5515014-5515014 on Build GRCh38
rs756268082 Chr.17: 5514885-5514885 on Build GRCh38
rs376646251 Chr.17: 5514899-5514899 on Build GRCh38
rs777520241 Chr.17: 5515019-5515019 on Build GRCh38
rs761021425 Chr.17: 5514977-5514977 on Build GRCh38
rs181358292 Chr.17: 5515071-5515071 on Build GRCh38
rs747817334 Chr.17: 5514918-5514918 on Build GRCh38
rs201878224 Chr.17: 5514916-5514916 on Build GRCh38
rs770497368 Chr.17: 5515042-5515042 on Build GRCh38
rs776946991 Chr.17: 5515076-5515076 on Build GRCh38
rs748851634 Chr.17: 5515036-5515036 on Build GRCh38
rs200037697 Chr.17: 5514937-5514937 on Build GRCh38
rs200752257 Chr.17: 5514913-5514913 on Build GRCh38
rs773267414 Chr.17: 5514931-5514932 on Build GRCh38
rs200829556 Chr.17: 5515069-5515069 on Build GRCh38
rs188604617 Chr.17: 5515052-5515052 on Build GRCh38
rs769506154 Chr.17: 5514921-5514921 on Build GRCh38
rs184023870 Chr.17: 5514896-5514896 on Build GRCh38
rs775762000 Chr.17: 5514968-5514968 on Build GRCh38
rs200833136 Chr.17: 5514985-5514985 on Build GRCh38
rs202212177 Chr.17: 5515002-5515002 on Build GRCh38
rs374010584 Chr.17: 5514873-5514873 on Build GRCh38
rs755841500 Chr.17: 5515016-5515016 on Build GRCh38
rs752779651 Chr.17: 5514991-5514991 on Build GRCh38
rs754014640 Chr.17: 5514897-5514897 on Build GRCh38
rs767877420 Chr.17: 5514964-5514964 on Build GRCh38
rs368144465 Chr.17: 5514874-5514874 on Build GRCh38
rs200352367 Chr.17: 5514992-5514992 on Build GRCh38
rs746893493 Chr.17: 5514914-5514914 on Build GRCh38
rs868641158 Chr.17: 5515020-5515020 on Build GRCh38
rs539985574 Chr.17: 5515004-5515004 on Build GRCh38
rs774952719 Chr.17: 5514956-5514956 on Build GRCh38
rs201588311 Chr.17: 5515001-5515001 on Build GRCh38
rs752934168 Chr.17: 5514884-5514884 on Build GRCh38
rs200001042 Chr.17: 5514883-5514883 on Build GRCh38
rs201238497 Chr.17: 5515049-5515049 on Build GRCh38
rs199776702 Chr.17: 5515054-5515054 on Build GRCh38
rs201167590 Chr.17: 5514863-5514863 on Build GRCh38
rs202032721 Chr.17: 5514989-5514989 on Build GRCh38
rs781139825 Chr.17: 5515012-5515012 on Build GRCh38
rs377020538 Chr.17: 5514923-5514923 on Build GRCh38
rs199842944 Chr.17: 5515048-5515048 on Build GRCh38
rs759983224 Chr.17: 5514957-5514957 on Build GRCh38
rs754905195 Chr.17: 5515006-5515006 on Build GRCh38

View Full Product Details